Product Image
Applied Biosystems™

TaqMan™ Respiratory Tract Microbiota Amplification Control

TaqMan Respiratory Tract Microbiota Amplification Control contains a linearized multi-target plasmid with target sequences for each available respiratory tract microbiota profiling assay.
Have Questions?
Catalog NumberQuantity
A391785 x 50 μL
Catalog number A39178
Price (MXN)
-
Quantity:
5 x 50 μL

TaqMan Respiratory Tract Microbiota Amplification Control contains a linearized multi-target plasmid with target sequences for each available respiratory tract microbiota profiling assay. The plasmid also contains target sequences for TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control, TaqMan Universal Extraction Control Organism (B. atrophaeus), and the human RNase P RPPH1 gene. The control can be included in respiratory tract microbiota profiling experiments as a positive control for panel-specific amplification.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormatTube(s)
No. of Reactions100 Reactions
Product LineTaqMan
Quantity5 x 50 μL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Volume250 μL
Concentration1 x 105 Copies/μL
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -25°C to -15°C

Frequently asked questions (FAQs)

What targets are in the TaqMan Respiratory Tract Microbiota Amplification Control?

The TaqMan Respiratory Tract Microbiota Amplification Control contains all the assays shown in Tables 1 and 2, page 7-8 and page 15 of the Respiratory Tract Microbiota Profiling Experiments v2 Application Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019506_RespiratoryTractMicrobiotaProfiling_OAv2_AG.pdf).