B-27™ Plus Neuronal Culture System
Gibco™
B-27™ Plus Neuronal Culture System
Improve the physiological relevance of neuronal cultures without sacrificing long-term health and stability. The B-27 Plus Neuronal Culture System with Neurobasal PhysioPlus Medium supports earlier, sustained neuronal activity, and exceptional cell viability to help accelerate scientific discoveries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
A40008171Neurobasal PhysioPlus Medium
A40008173Neurobasal PhysioPlus Medium (minus phenol red)
A3653401Neurobasal Plus Medium
Catalog number A40008171
Price (MXN)
-
Includes:
Neurobasal PhysioPlus Medium
The B-27 Plus Neuronal Culture System combines B-27 Plus Supplement with the advanced Neurobasal PhysioPlus Medium to support healthy, functional neuronal cultures. Designed for primary and hPSC-derived neurons, the system allows a more physiologically relevant culture environment that promotes accelerated neuronal maturation, dependable neuronal activity, and sustained viability over extended culture periods. The system is suitable across neurobiology applications from functional assays including MEA and live-cell imaging to high-content imaging and high-throughput plate-reader assays.
Key Features & Benefits
Morphological and synaptic maturation
  • Improves neurite outgrowth, synapse formation, neuronal morphology compared with conventional neuronal media approaches
  • Accelerates neurite outgrowth and increases the number of synaptic-positive puncta
Supports earlier and sustained neuronal activity
Obtaining neurons that survive, mature, and exhibit dependable functional activity in vitro remains a major challenge in neuroscience research. The B-27 Plus Neuronal Culture System was developed to support neuronal activity that arises earlier in culture and remains stable across extended timepoints, helping researchers obtain results faster and perform longitudinal studies with greater confidence. Product performance has been verified by whole-cell patch clamp recordings and MEA experiments.
The system is optimized to support:
  • Spontaneous neuronal activity
  • Formation of functional neuronal networks
  • Sustained electrophysiological activity over multiple weeks suitable for MEA workflows
More physiologically relevant neuronal culture conditions
Neurobasal PhysioPlus Medium is formulated to better reflect key aspects of the central nervous system (CNS) extracellular environment while maintaining robust neuronal health and performance.
  • Osmolarity consistent with CNS extracellular conditions
  • Reduced glucose concentrations, closer to physiological range
  • Neuroactive components at relevant concentrations
  • Verified electrophysiological properties of neurons in medium
Maintain healthy neurons longer in culture
The B-27 Plus Neuronal Culture System supports long-term neuron survival and health, enabling the highest survival rate of human stem cell-derived neurons and primary rodent neurons compared to other commercially available serum-free neuronal media systems. For example, 87% greater viability of rat cortical neurons was achieved with the B-27 Plus Neuronal Culture System compared to a leading competitor system, allowing your precious cell source to be seeded at lower density, seed more wells, and generate more data.
Researchers can culture neurons for extended periods to enable:
  • Neuronal maturation studies
  • Chronic treatment paradigms
  • Disease modeling applications
  • Longitudinal imaging experiments
  • Repeated functional measurements
Multiple basal medium options available to meet your needs
Neurobasal PhysioPlus Medium - the recommended basal medium, formulated to better replicate the CNS extracellular environment, supporting sustained neuronal activity, improved viability, and enhanced maturity over extended culture periods.
Neurobasal PhysioPlus Medium without phenol red – for use during analysis of neurons in imaging-sensitive applications or where the presence of phenol red is undesired.
Neurobasal Plus Medium – an alternative basal medium with a low osmolality, similar to the classic Neurobasal Medium (Cat. No. 21103049)
Built on a trusted neuronal culture foundation
The original B-27 Plus Neuronal Culture System expanded upon the industry-standard Neurobasal and B-27 platform to provide enhanced neuronal robustness and culture performance. The addition of Neurobasal PhysioPlus Medium represents the next evolution of this system, helping researchers generate healthier, more functional, and more physiologically relevant neuronal cultures for advanced neuroscience research.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Glucose ConcentrationReduced
IncludesNeurobasal PhysioPlus Medium
Product LineB-27, Neurobasal
Product TypeNeuronal Culture System
Quantity1 System
Shelf Life12 Months From Date of Manufacture
Shipping ConditionSupplement: Dry Ice, Basal: Ambient
ClassificationSerum-free
Culture TypeNeuronal Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesPhenol Red
Without AdditivesNone
Unit SizeEach
Contents & Storage
• 10 mL B-27 Plus Supplement (50X), store at -5°C to -20°C, protect from light
• 500 mL Neurobasal PhysioPlus Medium, store at 2°C to 8°C, protect from light
Frequently asked questions (FAQs)
What is the shelf-life of Gibco Neurobasal PhysioPlus Medium and how long can complete medium be stored?
Gibco Neurobasal PhysioPlus Medium has a shelf life of 12 months from the date of manufacture when stored at 4°C and protected from light. After supplementation with Gibco B-27 Plus Supplement, store complete medium at 4°C protected from light and use within 2-4 weeks.
How much glucose and Gibco GlutaMAX Supplement are in Gibco Neurobasal PhysioPlus Medium?
Gibco Neurobasal PhysioPlus Medium contains 6 mM glucose and 0.5 mM Gibco GlutaMAX Supplement.
Which Neurobasal medium should I choose based on glucose concentration?
Gibco Neurobasal PhysioPlus Medium is formulated with a reduced glucose concentration to more closely approximate physiological brain extracellular levels. Researchers requiring higher glucose concentrations may prefer other Neurobasal formulations depending on their application. If a medium completely free of glucose is needed, we offer Gibco Neurobasal-A Medium, no D-glucose, no sodium pyruvate.
Which supplement should I use with Gibco Neurobasal PhysioPlus Medium?
Use Gibco B-27 Plus Supplement with Gibco Neurobasal PhysioPlus Medium.
Do I need to gradually adapt neuronal cultures to Gibco Neurobasal PhysioPlus Medium?
Gradual adaptation to Gibco Neurobasal PhysioPlus Medium is typically not required. You can transition during a routine media change by replacing half the media volume with Neurobasal PhysioPlus Medium. We recommend verifying performance in your specific neuronal model when modifying an established protocol.