B-27™ Plus Neuronal Culture System with Neurobasal™ PhysioPlus Medium, reduced glucose, no additives - FAQs
B-27™ Plus Neuronal Culture System with Neurobasal™ PhysioPlus Medium, reduced glucose, no additives - FAQs
View additional product information for B-27™ Plus Neuronal Culture System - FAQs (A3653401, A40008173, A40008171)
9 product FAQs found
What is the shelf-life of Gibco Neurobasal PhysioPlus Medium and how long can complete medium be stored?
Gibco Neurobasal PhysioPlus Medium has a shelf life of 12 months from the date of manufacture when stored at 4°C and protected from light. After supplementation with Gibco B-27 Plus Supplement, store complete medium at 4°C protected from light and use within 2-4 weeks.
How much glucose and Gibco GlutaMAX Supplement are in Gibco Neurobasal PhysioPlus Medium?
Gibco Neurobasal PhysioPlus Medium contains 6 mM glucose and 0.5 mM Gibco GlutaMAX Supplement.
Which Neurobasal medium should I choose based on glucose concentration?
Gibco Neurobasal PhysioPlus Medium is formulated with a reduced glucose concentration to more closely approximate physiological brain extracellular levels. Researchers requiring higher glucose concentrations may prefer other Neurobasal formulations depending on their application. If a medium completely free of glucose is needed, we offer Gibco Neurobasal-A Medium, no D-glucose, no sodium pyruvate.
Which supplement should I use with Gibco Neurobasal PhysioPlus Medium?
Use Gibco B-27 Plus Supplement with Gibco Neurobasal PhysioPlus Medium.
Do I need to gradually adapt neuronal cultures to Gibco Neurobasal PhysioPlus Medium?
Gradual adaptation to Gibco Neurobasal PhysioPlus Medium is typically not required. You can transition during a routine media change by replacing half the media volume with Neurobasal PhysioPlus Medium. We recommend verifying performance in your specific neuronal model when modifying an established protocol.
When should I switch to Gibco Neurobasal PhysioPlus Medium during neuronal differentiation?
Use Gibco Neurobasal PhysioPlus Medium for neuronal maintenance and maturation after differentiation is complete. Continue your established differentiation protocol, then transition to Neurobasal PhysioPlus Medium with B-27 Plus Supplement when the culture consists predominantly of post-mitotic neurons displaying neuronal morphology and active differentiation has largely completed.
Can Gibco Neurobasal PhysioPlus Medium be used for neuronal organoids and assembloids?
Yes. Gibco Neurobasal PhysioPlus Medium can be used with three-dimensional neuronal models, including organoids and assembloids. We recommend introducing Neurobasal PhysioPlus Medium with B-27 Plus Supplement during maintenance, after differentiation is complete, and optimizing conditions for the specific model.
Can Gibco Neurobasal PhysioPlus Medium be used for neuron and glial cell co-culture?
Yes. Gibco Neurobasal PhysioPlus Medium can be used for co-culture of neurons and glial cells. Optimization may be needed based on the additional cell types and their media requirements. We recommend verifying conditions for each specific co-culture model.
Can Gibco Neurobasal PhysioPlus Medium be used with primary rodent neurons and iPSC-derived neurons?
Yes. Gibco Neurobasal PhysioPlus Medium has been demonstrated to support primary rodent neurons and multiple human PSC-derived neuronal models. Separate user manuals are available for primary neuron and iPSC-derived neuron applications.