Take your neuronal culture system further. Pair B-27™ Plus Supplement with the new Neurobasal™ PhysioPlus Medium to support robust, functionally active neuronal cultures with greater physiological relevance.
Neurobasal™ PhysioPlus Medium
Gibco™
Neurobasal™ PhysioPlus Medium
Optimized to facilitate consistent activity, exceptional viability, and long-term maturity in a physiologically relevant system, Neurobasal PhysioPlus Medium is recommended for culturing primary and hPSC-derived neurons.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWithout Additives
A4000817401No Phenol Red
A4000817201None
Catalog number A4000817401
Price (EUR)
115,44
Each
Without Additives:
No Phenol Red
Price (EUR)
115,44
Each
Neurobasal PhysioPlus Medium is an advanced cell culture medium for maturation and maintenance of primary and hPSC-derived neurons. The medium better replicates the central nervous system (CNS) extracellular environment and supports reliable neuronal activity that arises earlier in cultures and is sustained over long term culture durations. The medium is recommended in combination with B-27 Plus Supplement and helps provide consistent neural activity while maintaining exceptional cell viability, ensuring dependable experimental results in a physiologically relevant system.
Key Features & Benefits
Supports robust performance in functional assays
Neurobasal PhysioPlus Medium is optimized to support reliable neuronal activity and functional maturation across a wide range of neurobiology workflows, including multielectrode array (MEA) and calcium imaging applications.
  • Promotes earlier onset of spontaneous neuronal activity compared to traditional neuronal cell culture media
  • Enables a higher proportion of electrically active neurons across wells and plates
  • Supports sustained and stable network activity during long-term culture, increasing the useful experimental window of your neuronal cultures
Designed to better reflect the physiological CNS environment
Unlike conventional neuronal culture media optimized primarily for survival, Neurobasal PhysioPlus Medium was formulated to more closely mimic aspects of the extracellular environment of the central nervous system.
  • Physiologically informed formulation designed to support more in vivo-like neuronal behavior and help improve biological relevance of in vitro neuronal models
  • Optimized glucose concentration
  • Optimized neuroactive amino acid composition
  • Osmolarity aligned more closely with cerebrospinal fluid (CSF)
  • Supports electrophysiological properties, such as resting membrane potentials, spontaneous action potentials, and rheobase, more representative of native neuronal physiology, verified using whole-cell patch clamp recordings
Supports neuronal health, maturation, and long-term culture maintenance
Neurobasal PhysioPlus Medium was developed to support both neuronal function and cell health throughout extended culture durations.
  • Optimized nutrient composition supports sustained neuronal viability without compromising activity
  • Enables maintenance of healthy neuronal cultures for longer maturation and extended experimental timelines, including studies requiring more than a month in culture
  • Promotes extensive neurite outgrowth and neuronal network development
Optimized for modern neurobiology workflows
Neurobasal PhysioPlus Medium supports a broad range of neuronal cell culture applications and is recommended for use with B-27 Plus Supplement for optimal performance.
  • Compatible with both hPSC-derived neuronal models and primary rodent neurons
  • Supports applications including disease modeling, neurotoxicity studies, neuronal maturation, and functional screening
  • Available with or without phenol red to support imaging-based workflows
  • Enables researchers to generate more consistent and physiologically relevant experimental results
Built on the trusted Neurobasal legacy
Neurobasal PhysioPlus Medium builds upon the established Neurobasal platform trusted by neuroscientists for more than 30 years, delivering enhanced functional performance while maintaining the reliability and usability researchers expect from Gibco neuronal culture systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Glucose ConcentrationReduced
Product LineNeurobasal
Product TypeCell Culture Medium
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
Tested ForEndotoxin, Performance, Sterility, Osmolality, pH
ClassificationSerum-free
Culture TypeNeuronal Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesNone
Without AdditivesNo Phenol Red
Unit SizeEach
Frequently asked questions (FAQs)
What is the shelf-life of Gibco Neurobasal PhysioPlus Medium and how long can complete medium be stored?
Gibco Neurobasal PhysioPlus Medium has a shelf life of 12 months from the date of manufacture when stored at 4°C and protected from light. After supplementation with Gibco B-27 Plus Supplement, store complete medium at 4°C protected from light and use within 2-4 weeks.
How much glucose and Gibco GlutaMAX Supplement are in Gibco Neurobasal PhysioPlus Medium?
Gibco Neurobasal PhysioPlus Medium contains 6 mM glucose and 0.5 mM Gibco GlutaMAX Supplement.
Which Neurobasal medium should I choose based on glucose concentration?
Gibco Neurobasal PhysioPlus Medium is formulated with a reduced glucose concentration to more closely approximate physiological brain extracellular levels. Researchers requiring higher glucose concentrations may prefer other Neurobasal formulations depending on their application. If a medium completely free of glucose is needed, we offer Gibco Neurobasal-A Medium, no D-glucose, no sodium pyruvate.
Which supplement should I use with Gibco Neurobasal PhysioPlus Medium?
Use Gibco B-27 Plus Supplement with Gibco Neurobasal PhysioPlus Medium.
Do I need to gradually adapt neuronal cultures to Gibco Neurobasal PhysioPlus Medium?
Gradual adaptation to Gibco Neurobasal PhysioPlus Medium is typically not required. You can transition during a routine media change by replacing half the media volume with Neurobasal PhysioPlus Medium. We recommend verifying performance in your specific neuronal model when modifying an established protocol.