Neurobasal™ PhysioPlus Medium, minus phenol red - FAQs
Neurobasal™ PhysioPlus Medium, minus phenol red - FAQs
View additional product information for Neurobasal™ PhysioPlus Medium - FAQs (A4000817201, A4000817401)
This product does not have any FAQs available.