What is included in Genexus Barcodes 1-32 HD (Cat. No. A40261)?
Genexus Barcodes 1-32 HD provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes HD 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.