Genexus™ Barcodes 1-32 HD
Ion Torrent™
Genexus™ Barcodes 1-32 HD
The Ion Torrent Genexus Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered withMás información
Have Questions?
Número de catálogoCantidad
A4026132 Reactions
Número de catálogo A40261
Precio (MXN)
7,925.58
Each
Cantidad:
32 Reactions
Precio (MXN)
7,925.58
Each
The Ion Torrent Genexus Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. The barcodes can be flexibly utilized in increments of 1 to 32, until all barcodes are exhausted.When used in conjunction with Ion Torrent Genexus Library Strips 1 and 2-HD, this kit supports flexible, fully automated preparation and normalization of up to 32 Ion AmpliSeq HD libraries. For Ion AmpliSeq-based assays, choose from other available barcode kits: Ion Torrent Genexus Barcodes 1-32 AS, 33-64 AS, 65-96 AS, or 1-96 AS.
As part of the Genexus Integrated Sequencer workflow, the Genexus Barcodes 1-32 HD kit:
• Automates library preparation for 200–400 base-read Ion AmpliSeq HD libraries
• Supports intake variability of clinical research samples in a cost-effective manner
• Maximizes flexibility with the ability prepare 1 to 32 single-pool libraries per run
• Enables parallel processing of up to four compatible assays in a single run
• Delivers fast and simple instrument setup, with less than five minutes of hands-on time
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)Secuenciación
Para utilizar con (equipo)Genexus™ Integrated Sequencer
BibliotecasTargeted Sequencing Library
N.º de reacciones32
Línea de productosGenexus
Tipo de productoBarcode
Cantidad32 Reactions
Tipo de muestraDNA, RNA
Condiciones de envíoTemperatura ambiente
Paso del flujo de trabajoLibrary Generation
Sequencing TypeGenome, DNA, and RNA Sequencing
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente.
Preguntas frecuentes
What is included in Genexus Barcodes 1-32 HD (Cat. No. A40261)?
Genexus Barcodes 1-32 HD provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes HD 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Will the Genexus Software notify me if the barcodes had been previously used?
When necessary, the software will alert the user during the run plan setup that not enough barcodes are available.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How long can the barcodes remain on the Genexus platform?
Barcodes plates can remain on the instrument deck until all barcodes are used. Once you remove the barcode plate, you cannot put it back on to use again as the instrument has no way of tracking barcode usage.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.