Genexus™ Primer Pool Tubes
Ion Torrent™

Genexus™ Primer Pool Tubes

The Ion Torrent Genexus Primer Pool Tubes kit contains 50 empty assemblies of the custom tubes required to deliver primer oligos into the automated library preparation reaction.
Have Questions?
Catalog NumberQuantity
A40262200 Reactions
Catalog number A40262
Price (MXN)
10,205.93
Each
Quantity:
200 Reactions
Price (MXN)
10,205.93
Each

The Ion Torrent Genexus Primer Pool Tubes kit contains 50 empty assemblies of the custom tubes required to deliver primer oligos into the automated library preparation reaction. Each assembly is serialized for traceability and can be used with Ion AmpliSeq and Ion AmpliSeq HD-based assays. Pipette an aliquot of panel primers from a custom or AmpliSeq-on-Demand panel in order to run the assay on the Genexus Integrated Sequencer.

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Integrated Sequencer
LibrariesTargeted Sequencing Library
No. of Reactions200
Product LineGenexus
Product TypePrimer Pool Tube
Quantity200 Reactions
Sample TypeDNA, RNA
Shipping ConditionRoom Temperature
Workflow StepLibrary Generation
Sequencing TypeGenome, DNA, and RNA Sequencing
Unit SizeEach

Frequently asked questions (FAQs)

How many samples of Oncomine Precision Assay GX can I run on each lane of the GX5 Chip?

The capacity of the GX5 Chip varies depending on the sample type:
- For plasma cfTNA samples, the workflow is configured to accommodate 1 sample per GX5 Chip lane (or 4 samples on a single GX5 Chip).
- For FFPE DNA+RNA samples, the workflow is configured to accommodate 4 samples per GX5 Chip lane.
- For FFPE DNA-only or RNA-only samples, the workflow is configured to accommodate 8 samples per GX5 Chip lane.

How many sequencing reactions can I perform with Oncomine Precision Assay GX (Cat. No. A44350)?

The maximum number of samples that can be analyzed per kit depends upon the type of analysis that you want to perform. Each kit provides the targeted pancancer panel and library reagents sufficient to perform up to 32 sequencing reactions on the Genexus Integrated Sequencer.

- Simultaneous DNA- and RNA-based variant analysis for up to 16 FFPE tissue samples (one DNA reaction and one RNA reaction per sample)
- DNA-only or RNA-only variant analysis for up to 32 FFPE tissue samples
- Cell-free TNA (cfTNA) variant analysis (combined DNA and RNA) for up to 32 plasma samples

How much of each sample do I need for the Oncomine Precision Assay GX?

For solid tumor samples, the optimal input amount is 10 ng with validated input in the range of 5-15 ng. For liquid biopsy samples, the optimal input amount is 20 ng with validated input in the range of 5-40 ng.

For optimal results, load 20 µL of input with a concentration of ≥0.67 ng/µL for FFPE DNA and FFPE RNA samples and 20 µL of input with a concentration of ≥1.33 ng/µL for cfTNA samples. If the concentration of the sample is lower than optimal, use as much input as possible for each reaction.

What controls should I use with Oncomine Precision Assay GX?

Here are the various controls we recommend depending upon the starting material:

FFPE DNA samples
- Structural Multiplex Reference Standard (FFPE), Cat. No. HD789 (Horizon™)
- EGFR Gene-Specific Multiplex Reference Standard (FFPE) 5% AF, Cat. No. HD300 (Horizon™)
- KRAS Gene-Specific Multiplex Reference Standard (FFPE), Cat. No. HD301 (Horizon™)
- Seraseq® Lung & Brain CNV Mix, + 3 copies, Cat. No. 0710-0414 (SeraCare®)
- Seraseq® Breast CNV Mix, + 6 copies, Cat. No. 0710-0412 (SeraCare®)

FFPE RNA samples
- Seraseq® FFPE Tumor Fusion RNA v4 Reference Material, Cat. No. 0710-0496 (SeraCare®)
- Seraseq® FFPE NTRK Fusion RNA Reference Material, Cat. No. 0710-1031 (SeraCare®)

cfTNA samples
- Multiplex I cfDNA Reference Standard Set, Cat. No. HD780 (Horizon™)

Which sample types can I use with Oncomine Precision Assay GX?

Oncomine Precision Assay GX is compatible with solid tumor samples (FFPE DNA or FFPE RNA) as well as liquid biopsy samples (cfTNA).