Genexus™ Templating Strips 3-GX5™ and 4
Ion Torrent™
Genexus™ Templating Strips 3-GX5™ and 4
The Ion Torrent Genexus Templating Strips 3-GX5 and 4 kit provides eight each of the strips required to perform automatedMás información
Have Questions?
Número de catálogoCantidad
A402632 Runs
Número de catálogo A40263
Precio (MXN)
50,147.08
Each
Cantidad:
2 Runs
Precio (MXN)
50,147.08
Each
The Ion Torrent Genexus Templating Strips 3-GX5 and 4 kit provides eight each of the strips required to perform automated template preparation and chip loading of two Ion Torrent GX5 chips using the Ion Torrent Genexus Integrated Sequencer. This kit, combined with the GX5 Chip and Genexus Coupler, supports flexible, fully automated template preparation and chip loading of libraries prepared using the Genexus Integrated Sequencer or with an alternative protocol.
As part of the Genexus Integrated Sequencer workflow, the Genexus Templating Strips 3-GX5 and 4 kit:
  • Automates template preparation and GX5 Chip loading for 200–400 base-read Ion AmpliSeq or Ion AmpliSeq HD libraries
  • Supports intake variability of clinical research samples in a cost-effective manner
  • Maximizes flexibility with the ability to load one to four chip lanes per run
  • Enables parallel processing of up to four compatible assays in a single run
  • Delivers fast and simple instrument setup, with less than five minutes of hands-on time
The Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)Secuenciación
Línea de productosGenexus
Tipo de productoTemplating Strip
Cantidad2 Runs
Condiciones de envíoAprobado para su envío en hielo húmedo o seco
Unit SizeEach
Contenido y almacenamiento
• Genexus Strip 4, store at -10 to -30°C
• Genexus Strip 3-GX5, store at 2–8°C
Preguntas frecuentes
What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?
Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.
Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?
Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:
Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784
Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01
What are the sample input requirements for Oncomine Myeloid Research Assay GX?
The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).
What sample controls should I use with the Oncomine Myeloid Research Assay GX?
The recommended controls are:
DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056
RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)
How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?
Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).