Genexus™ Sequencing Kit
Ion Torrent™
Genexus™ Sequencing Kit
Have Questions?
Número do catálogoQuantity
A402712 Runs
Número do catálogo A40271
Preço (BRL)
-
Quantity:
2 Runs
The Ion Torrent Genexus Sequencing Kit provides the materials required to perform two initializations and automated sequencing of two multilane chips using the Ion Torrent Genexus Integrated Sequencer. These innovative sequencing consumables are stable on the instrument for up to two weeks after initialization, allowing flexibility to use the chip lanes according to sample throughput needs. Genexus Bottles 1, 2, and 3 are shelf-stable and the Genexus Cartridge has a minimized footprint, limiting the required storage space at -20°C.
As part of the Genexus Integrated Sequencer workflow, the Genexus Sequencing Kit:
• Automates sequencing for 200–400 base-read Ion AmpliSeq or Ion AmpliSeq HD libraries
• Supports intake variability of clinical research samples in a cost-effective manner
• Maximizes flexibility with the ability to sequence one to four chip lanes per run
• Enables parallel processing of up to four compatible assays in a single run
• Delivers fast and simple instrument setup, with less than five minutes of hands-on time
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
For Use With (Application)Comparative Sequencing (Somatic Mutations - 10:50)
For Use With (Equipment)Genexus™ Integrated Sequencer
Product LineGenexus
Product TypeSequencing Kit
Quantity2 Runs
Shipping ConditionQualified for Ambient or Wet Ice
FormatKit
Unit SizeEach
Conteúdo e armazenamento
• Genexus Cartridge, store at -10 to -30°C
• Genexus Bottle 2, store at room temperature
• Genexus Bottles 1 and 3, store at room temperature
Frequently asked questions (FAQs)
What is the purpose of the cartridge in the Genexus Sequencing Kit Combo?
The Genexus Cartridge contains key sequencing consumables used by the Genexus Integrated Sequencer to perform automated sequencing runs. In short, it is the main sequencing reagent cartridge required for instrument initialization and chip sequencing.
How many boxes of the Genexus Sequencing Kit Combo do I need to run 24 lanes?
You would need 3 kits. Each A40271 kit supports up to 8 lanes total (2 full 4-lane chips), so 24 lanes require 3 kits.
Is there an SDS available for the Genexus Sequencing Kit Combo?
Yes. The product page includes SDS/documents access for the Genexus Sequencing Kit, so an SDS should be available through the Thermo Fisher Documents and Certificates section.
Is the Genexus Sequencing Kit Combo compatible with the Ion GeneStudio S5 systems?
No. The Genexus Sequencing Kit (A40271) is intended for use with the Genexus Integrated Sequencer, not the Ion GeneStudio S5 system.
What are the correct shipping conditions for the Genexus Cartridges that are a part of the Genexus Sequencing Kit (Cat. No. A40271) and Genexus GX5 Starter Packs (Cat. Nos. A40279, A40280)?
The Genexus Cartridge should be shipped on supergels at -15 to -50 degrees C, in a polystyrene box. If the cartridges are shipped with dry ice or arrive thawed, they should not be used, and replacement would need to be issued.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.