Genexus™ GX5™ Starter Pack-HD
Genexus™ GX5™ Starter Pack-HD
Ion Torrent™

Genexus™ GX5™ Starter Pack-HD

The Ion Torrent Genexus GX5 Starter Pack for Ion AmpliSeq HD panels provides one set of all core consumables requiredRead more
Have Questions?
Catalog NumberQuantity
A4028032 Reactions
Catalog number A40280
Price (USD)
13,420.00
Each
Add to cart
Quantity:
32 Reactions
Price (USD)
13,420.00
Each
Add to cart
The Ion Torrent Genexus GX5 Starter Pack for Ion AmpliSeq HD panels provides one set of all core consumables required to run the Ion Torrent Genexus Integrated Sequencer. This starter pack supports flexible, fully automated preparation and normalization of up to 32 Ion AmpliSeq HD libraries, as well as sequencing for up to eight chip lanes (two full chips). The Ion Torrent GX5 Chip is used in conjunction with this starter pack and must be ordered separately.

As part of the Genexus Integrated Sequencer GX5 workflow, the Genexus GX5 Starter Pack-HD:
• Automates library preparation, templating, and sequencing for 200–400 base-read Ion AmpliSeq HD libraries
• Supports intake variability of clinical research samples in a cost-effective manner
• Maximizes flexibility with the ability to sequence one to four chip lanes per run
• Enables parallel processing of up to four compatible assays in a single run
• Delivers fast and simple instrument set-up, with less than five minutes of hands-on time

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Comparative Sequencing (Somatic Mutations - 10:50)
For Use With (Equipment)Genexus Integrated Sequencer
No. of Reactions32
Product LineGX5, Genexus
Product TypeStarter Pack-AS
Quantity32 Reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Unit SizeEach
Contents & Storage
1 each of:
• Genexus Strip 2-HD, store at -10 to -20°C
• Genexus Strip 1, store at 2–8°C
• Genexus Barcodes 1-32 HD, store at room temperature
• Genexus Strip 4, store at -10 to -20°C
• Genexus Strip 3-GX5, store at 2–8°C
• Genexus Pipette Tips, store at room temperature
• Genexus Cartridge, store at -10 to -20°C
• Genexus Bottle 2, store at room temperature
• Genexus Bottles 1 and 3, store at room temperature

Frequently asked questions (FAQs)

What are the correct shipping conditions for the Genexus Cartridges that are a part of the Genexus Sequencing Kit (Cat. No. A40271) and Genexus GX5 Starter Packs (Cat. Nos. A40279, A40280)?

The Genexus Cartridge should be shipped on supergels at -15 to -50 degrees C, in a polystyrene box. If the cartridges are shipped with dry ice or arrive thawed, they should not be used, and replacement would need to be issued.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the components of the Genexus GX5 Starter Pack-HD?

Genexus GX5 Starter Pack-HD (Cat. No. A40280) supplies the following components for Ion AmpliSeq HD library preparation and sequencing using a Genexus-ready assay:
- Genexus Strip 1, 8 strips
- Genexus Strip 2‑HD, 8 strips
- Genexus Strip 3‑GX5, 8 strips
- Genexus Strip 4, 8 strips
- Genexus Barcodes 1-32 HD, 1 plate
- Genexus Pipette Tips, 12 racks
- Genexus Cartridge, 2 cartridges
- Genexus Bottle 2, 4 bottles
- Genexus Bottles 1 and 3, 2 bottles each
Note:
- Genexus Barcodes 1-32 HD (Cat. No. A40261) and Genexus Pipette Tips (Cat. No. A40266) are also orderable individually.
- Library and template strips are orderable separately in paired sets as Genexus Library Strips 1 and 2-HD (Cat. No. A40255) and Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263).
- The Genexus Cartridge, Genexus Bottle 2, and Genexus Bottles 1 and 3 are orderable as a set in the Genexus Sequencing Kit (Cat. No. A40271).
- For custom assays, Genexus Primer Pool Tubes (Cat. No. A40262) must be ordered separately.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.