Invitrogen™

PRKCA (PKC alpha) Recombinant Human Protein

Catalog number: A41892
Invitrogen™

PRKCA (PKC alpha) Recombinant Human Protein

Catalog number: A41892
Catalog Number
A41892
Unit Size
Price (USD)
1,660.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
7,000.00
Your Price:
Availability
***
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
484.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A41892
1,660.00
Your Price:
***
A418931 mg
7,000.00
Your Price:
***
A4189110 µg
484.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PKC family of serine/threonine kinases, including PRKCA (PKC alpha), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.

Specifications

Accession Number
NP_002728.1
Shipping Condition
Dry Ice
Expression System
Insect
Form
Liquid
Protein Form
Full Length, Active
Protein Subtype
Serine⁄Threonine Kinases
Research Category
Drug Development
Species
Human
Validated Application
Kinase Assay
Gene ID (Entrez)
PRKCA
Molecular Weight
110.0 kDa
Quantity
100 µg
Gene Alias
PKCA, PKC-A, PRKACA
KinaseFamily
PKC Kinase Family
Recombinant
Yes
Protein Tag
GST Tag

Contents & Storage

Storage Buffer: 50 mM Tris-HCl, pH 8.0, 100 mM NaCl, 5 mM DTT, 15 mM reduced glutathione, 20% glycerol

Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.

Figures

Frequently asked questions (FAQs)

Citations & References