MagMAX™ Viral/Pathogen Proteinase K - FAQs
MagMAX™ Viral/Pathogen Proteinase K - FAQs
View additional product information for MagMAX™ Viral/Pathogen Proteinase K - FAQs (A42363)
1 product FAQs found
What is the concentration of MagMAX Viral/Pathogen Proteinase K (Cat. No. A42363)?
The concentration of MagMAX Viral/Pathogen Proteinase K is proprietary.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.