MagMAX™ Viral/Pathogen Proteinase K
Applied Biosystems™

MagMAX™ Viral/Pathogen Proteinase K

This Proteinase K is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid IsolationRead more
Have Questions?
Catalog NumberQuantity
A4236310 mL
Catalog number A42363
Price (MXN)
14,361.40
Each
Quantity:
10 mL
Price (MXN)
14,361.40
Each
This Proteinase K is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more Proteinase K than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing (NGS)
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeViral/Pathogen Proteinase K
Quantity10 mL
Sample TypeLiquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
Shipping ConditionRoom Temperature
TargetViral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What is the concentration of MagMAX Viral/Pathogen Proteinase K (Cat. No. A42363)?

The concentration of MagMAX Viral/Pathogen Proteinase K is proprietary.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.