Thermal Printer for iBright and iGlow Imaging Systems
Thermal Printer for iBright and iGlow Imaging Systems
Thermal Printer for iBright and iGlow Imaging Systems

Thermal Printer for iBright and iGlow Imaging Systems

This high-speed and compact thermal printer is plug-and-play and connects directly to an iBright Imaging System.
Have Questions?
카탈로그 번호수량
A452351 printer
카탈로그 번호 A45235
제품 가격(KRW)
2,807,000
Each
수량:
1 printer
제품 가격(KRW)
2,807,000
Each

The Sony UP-X898MD Digital Graphic Printer is a thermal printer that supports direct printing from iBright and iGlow Imaging Systems. Capable of delivering high quality black and white prints in less than two seconds, this high-speed and compact printer is plug-and-play and connects directly to iBright and iGlow Imaging Systems through one of the available USB ports. Print high-quality images in seconds with no need for a network connection. Images in the view port print as displayed with current zoom level and analysis annotations preserved.

Features of the Digital Graphic Printer include:

  • High-speed printing—prints 320 mm x 100 mm images in approximately two seconds
  • Compact, space-saving design (88 mm x 154 mm x 240 mm (HxWxD))
  • High-density 325 dpi printing (1280 x 500 pixels)
  • 256 grey-scale levels
  • USB 2.0 interface
For Research Use Only. Not for use in diagnostic procedures.
사양
용량Up to 254
치수3.46 x 6.06 x 9.45 in (88 x 154 x 240 mm)
용도(장비)iBright and iGlow Imaging Systems
수량1 printer
분해능325 DPI
배송 조건Room Temperature
중량(영국식 단위)5.5 lb
중량(미터법)2.5 kg
용도(애플리케이션)Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
제품라인iBright
제품 유형Imaging System
Unit SizeEach

자주 묻는 질문(FAQ)

Does the iGlow Gel Documentation System support printing to a thermal printer?

Yes. The iGlow Gel Documentation System is compatible with the Sony UP-X898MD thermal printer (Cat. No. A45235). The printer connects directly to the imager using a USB A-B cable, allowing users to print images directly from the system.

How are images on an iBright Imaging System printed on a thermal printer?

Once communication is established between the IBright imager and the thermal printer, a print button will become available on the main screen near the image display. Pressing this button will initiate printing of the image currently shown on the instrument screen. This is a separate workflow from the print settings available through the system menu, which are used for printing image reports on a standard network printer.

For more information on iBright features and capabilities, visit this page.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How does the Thermal Printer for iBright Imaging Systems (Cat. No. A45235) interface with the iBright Imaging Systems?

To establish communication between the Thermal Printer and the iBright imager, first the communication channel on the printer needs to be updated. This can be done by accessing the printer menu, navigating from Digital to Driver and then using the up down toggle to set the printer to channel 897. Once that is complete, the thermal printer can then be connected directly to one of the iBright imager rear USB ports via A to B type USB cable. After installation, the thermal printer can be controlled directly with the iBright onboard software via the on-screen print functionality.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the weight of the Thermal Printer for iBright Imaging Systems?

The Thermal Printer for iBright Imaging Systems weighs approximately 5.5 lbs (2.5 kg).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the dimensions of the Thermal Printer for iBright Imaging Systems?

Here are the dimensions of the Thermal Printer for iBright Imaging Systems:
Approximately 3.46 x 6.06 x 9.45 inches (88 x 154 x 240 mm).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.