Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
Ion 510™ & Ion 520™ & Ion 530™ Kit for Agrigenomics – Chef (1 sequencing run per initialization)
Ion Torrent™

Ion 510™ & Ion 520™ & Ion 530™ Kit for Agrigenomics – Chef (1 sequencing run per initialization)

The Ion 510 & Ion 520 & Ion 530 Kit-Chef (1 sequencing run per initialization) enables robust and automated templateRead more
Have Questions?
Catalog NumberQuantity
A458498 Reactions
Catalog number A45849
Price (USD)
-
Quantity:
8 Reactions
The Ion 510 & Ion 520 & Ion 530 Kit-Chef (1 sequencing run per initialization) enables robust and automated template preparation and sequencing of up to 400 base-read libraries using the Ion Chef System and the Ion S5 or Ion S5 XL sequencing system. The Ion 510 & Ion 520 & Ion 530 Kit contains pre-packaged single-use template and sequencing reagent cartridges with integrated sample tracking, delivering an automated workflow for sequencing of 8 loaded chips with complete run traceability. This kit accommodates 1 sequencing run per initialization. For 2 sequencing runs per initialization (200 base-read applications only), see Cat. No. A44875.

The Ion 510 & Ion 520 & Ion 530 Kit-Chef workflow:
• Provides template preparation and chip loading reagents for up to 400 base-read libraries
• Simplifies your research with ready-to-use, disposable reagent cartridges for sequencing
• Enables fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
• Leverages the latest advancements in data quality and accuracy
• Maximizes flexibility with compatibility for the Ion 510, Ion 520, and Ion 530 chips

The Ion S5 and Ion S5 XL sequencers, along with the Ion Chef System, offer reproducible sequencing workflows, with minimal hands-on time. Choose to pair the Ion 510 & Ion 520 & Ion 530 Kit with the Ion 510 Chip Kit (up to 3 million reads per chip), the Ion 520 Chip Kit (up to 5 million reads per chip), or Ion 530 Chip Kit (up to 20 million reads), allowing for flexibility of total throughput. For even greater flexibility, combine different chip types in a single Ion Chef run when using Torrent Suite Software 5.6 and above. For de novo research applications, add the Ion S5 Calibration Standard to prepared libraries for improved base-calling accuracy.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)De Novo Sequencing - High Throughput (cDNA)
For Use With (Equipment)Ion Proton™ System
No. of Reactions8 reactions
Product LineIon Chef
Product TypeDNA Sequencing Kit
Quantity8 Reactions
Shipping ConditionWet Ice
FormatKit
Unit SizeEach
Contents & Storage
• Ion S5 Chef supplies, store at room temperature
• Ion S5 Chef solutions, store at room temperature
• Ion 510 & Ion 520 & Ion 530 Chef reagents, store at -5 to -30°C
• Ion S5 Sequencing solutions, store at room temperature
• Ion S5 Sequencing reagents, store at -5 to -30°C

Frequently asked questions (FAQs)

Why did my Ion Chef Instrument stop during a run?

- The instrument may have encountered an error. We recommend recording the error shown on the instrument display, then tapping OK. For further assistance, contact Technical Support at techsupport@thermofisher.com.
- A consumable may have been improperly loaded. We recommend ensuring that all consumables are correctly loaded. For instructions, see the user guide for your templating kit. For further assistance, contact Technical Support at techsupport@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Why is the door of the Ion Chef Instrument not opening?

- An obstruction may be present on or around the door mechanism. We recommend removing the obstruction blocking the door, then resuming normal instrument operation.
- The instument may be experiencing a hardware or software error. We recommend contacting Technical Support at techsupport@thermofisher.com.C8

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Why does the Ion Chef Instrument not display the home screen when powered on?

Multiple causes are possible. We recommend powering off the instrument, waiting for 30 secs and then powering on the instrument. If you continue to experience the issue, contact Technical Support at techsupport@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many runs per initialization can I perform on the Ion S5 and Ion S5 XL Systems, when using in conjunction with the Ion Chef?

You can perform one or two runs per initialization depending on the kit you have:

- Ion 510 & Ion 520 & Ion 530 Kit - Chef (1 sequencing run per initialization), Cat. No. A34019)

- Ion 510 & Ion 520 & Ion 530 Kit - Chef (2 sequencing runs per initialization), Cat. No. A34461)

- Ion 540 Kit - Chef (1 sequencing run per initialization), Cat. No. A27759

- Ion 540 Kit - Chef (2 sequencing runs per initialization), Cat. No. A30011

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.