What data output sizes should I expect from the Ion Torrent GX7 Chip?
Typical sequencing output files include approximately 40 GB for uBAM files, 45 GB for BAM files, and 25 MB for VCF files. The Ion Torrent GX7 Chip generates approximately 20–25 million sequencing reads per lane for 200-base-read libraries, corresponding to approximately 80–100 million reads when all four lanes of the chip are used. Refer to the relevant Genexus Software User Guide for full details on temporary and intermediate files.