1,8-ANS (1-Anilinonaphthalene-8-Sulfonic Acid), high purity
Invitrogen™

1,8-ANS (1-Anilinonaphthalene-8-Sulfonic Acid), high purity

A sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.

Have Questions?
Catalog NumberQuantity
A47100 mg
Catalog number A47
Price (USD)
317.65
Online exclusive
338.00
Save 20.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
317.65
Online exclusive
338.00
Save 20.35
Each

1,8-ANS (1-anilinonaphthalene- 8-sulfonic acid) is essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes. This property makes it a sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Quantity100 mg
Shipping ConditionRoom Temperature
Sub Cellular LocalizationCell Membranes & Lipids
ColorBlue
For Use With (Equipment)Fluorescence Microscope
Product Type1,8-ANS
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.

Citations & References (548)

Citations & References
Abstract
Conformational change in the pheromone-binding protein from Bombyx mori induced by pH and by interaction with membranes.
Authors:Wojtasek H,Leal WS
Journal:The Journal of biological chemistry
PubMed ID:10521490
Cofactor-induced refolding: refolding of molten globule carbonic anhydrase induced by Zn(II) and Co(II).
Authors:Andersson D,Hammarström P,Carlsson U
Journal:Biochemistry
PubMed ID:11258876
Rapid irreversible G protein alpha subunit misfolding due to intramolecular kinetic bottleneck that precedes Mg2+ "lock" after GTP/GDP exchange.
Authors:Zelent B,Veklich Y,Murray J,Parkes JH,Gibson S,Liebman PA
Journal:Biochemistry
PubMed ID:11583165
Stoichiometric exchange of GTP for GDP on heterotrimeric G protein alpha (Galpha) subunits is essential to most hormone and neurotransmitter initiated signal transduction. Galphas are stably activated in a Mg2+ complex with GTPgammaS, a nonhydrolyzable GTP analogue that is reported to bind Galpha, with very high affinity. Yet, it is ... More
Differential perturbation of intersubunit and interdomain communications by glycine 141 mutation in Escherichia coli CRP.
Authors:Cheng X, Lee JC
Journal:Biochemistry
PubMed ID:9425025
Upon binding of cAMP, concomitant changes in CRP structure across the subunit and domain interfaces are observed. In order to identify the structural elements involved in the coupling of interfacial interactions, structural perturbation was introduced at residue 141 by site-directed mutagenesis. Thermodynamic parameters defining protein stability, cAMP binding, and subunit ... More
Phosphorescence from 2-(p-toluidinyl)naphthalene-6-sulfonate and 1-anilinonaphthalene-8-sulfonate, commonly used fluorescence probes of biological structures.
Authors:Maniara G, Vanderkooi JM, Bloomgarden DC, Koloczek H
Journal:Photochem Photobiol
PubMed ID:3344289