TaqMan™ SCID/SMA Plus Assay
Applied Biosystems™

TaqMan™ SCID/SMA Plus Assay

A multiplex real-time PCR assay that detects the SMN1 gene, TREC, and KREC, as well as the RNase P gene as an internal genomic control.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A4856920,000 Reactions
A485661000 Reactions
A485674000 Reactions
A485688000 Reactions
Catalog number A48569
Price (MXN)
-
No. of Reactions:
20,000 Reactions

The Applied Biosystems TaqMan SCID/SMA Plus Assay is a multiplex real-time PCR assay that detects the SMN1 gene, TREC, and KREC, as well as the RNase P gene as an internal genomic control. For an assay that detects only SMN1 and TREC, please see the TaqMAN SCID/SMA Assay (Cat. No. A48566).

Features of the TaqMan SCID/SMA Plus Assay include:

  • Optimized for SCID and SMA
  • Direct extraction from DBS samples for reduced hands-on time and cost
  • Compatible with 96- or 384-reaction protocols
  • High-quality results for 96 samples within 55 min using the fast cycling protocol
  • Minimal liquid-dispensing steps for easy automation

Complete solution from DBS to result
The TaqMan SCID/SMA Plus Assay detects SMN1, TREC, and KREC, as well as the RNase P gene as an internal genomic control. The assay solution contains a mixture of primers and probes for each target plus an SMN2 blocker. From DBS to result, less than 40 min of hands-on time and only four reagent-dispensing steps are required for 96-well plates. One additional pipetting transfer step from 96- to 384-well plates is needed. A real-time PCR instrument with at least five filters is required to detect the reporter dyes plus the passive reference dye.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)QuantStudio™ 12 k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, QuantStudio™ Dx System (in RUO Mode)
No. of Reactions20,000 Reactions
Product LineTaqMan
Quantity20,000 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
• DNA Extract All Reagents Kit
• TaqPath ProAmp Multiplex Master Mix
• TaqMan SCID/SMA Plus Assay

Upon receipt, store at -20°C. After opening, store at 4°C.

Frequently asked questions (FAQs)

How can I order TREC and KREC plasmids directly from GeneArt for use with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?

Please refer to the following TaqMan SCID/SMA Plus Assay User Guide (pg 6) for instructions on ordering plasmids from GeneArt.

Are the TREC and KREC plasmids included when I purchase the TaqMan SCID/SMA Assay or TaqMan SCID/SMA Plus Assay?

No, you will need to order the TREC and KREC plasmids separately. Please refer to the following TaqMan SCID/SMA Plus Assay User Guide (pg 6) for instructions on ordering plasmids from GeneArt.

Are the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay compatible with QuantStudio 6 and 7 Flex Real-Time PCR instruments using QuantStudio Software v1.6.0 (or newer) and firmware v1.0.4 (or newer)?

Yes, the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are compatible with QuantStudio 6 and 7 instruments using QuantStudio Software v1.6.0 (or newer) and firmware v1.0.4 (or newer).

Who is responsible for QuantStudio instrument dye calibration for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay prior to an FAS onsite customer training?

The user is responsible for ordering the required calibration plates prior to the onsite training. If you purchase the onsite training, you may ask the FAS to complete the instrument dye calibration.

What is included when I purchase the Newborn Screening/NBS training (Cat. No. A48619) for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?

The NBS training (Cat. No. A48619) is a 1 day onsite FAS training which includes hands-on workflow training.