TaqMan™ SCID/SMA Plus Assay, 20,000 reactions - FAQs
TaqMan™ SCID/SMA Plus Assay, 20,000 reactions - FAQs
View additional product information for TaqMan™ SCID/SMA Plus Assay - FAQs (A48568, A48569, A48566, A48567)
50 product FAQs found
How can I order TREC and KREC plasmids directly from GeneArt for use with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
Please refer to the following TaqMan SCID/SMA Plus Assay User Guide (pg 6) for instructions on ordering plasmids from GeneArt.
Are the TREC and KREC plasmids included when I purchase the TaqMan SCID/SMA Assay or TaqMan SCID/SMA Plus Assay?
No, you will need to order the TREC and KREC plasmids separately. Please refer to the following TaqMan SCID/SMA Plus Assay User Guide (pg 6) for instructions on ordering plasmids from GeneArt.
Are the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay compatible with QuantStudio 6 and 7 Flex Real-Time PCR instruments using QuantStudio Software v1.6.0 (or newer) and firmware v1.0.4 (or newer)?
Yes, the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are compatible with QuantStudio 6 and 7 instruments using QuantStudio Software v1.6.0 (or newer) and firmware v1.0.4 (or newer).
Who is responsible for QuantStudio instrument dye calibration for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay prior to an FAS onsite customer training?
The user is responsible for ordering the required calibration plates prior to the onsite training. If you purchase the onsite training, you may ask the FAS to complete the instrument dye calibration.
What is included when I purchase the Newborn Screening/NBS training (Cat. No. A48619) for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
The NBS training (Cat. No. A48619) is a 1 day onsite FAS training which includes hands-on workflow training.
Do you recommend any analysis criteria for calling SCID with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
We cannot provide a specific diagnostic copy number cutoff for SCID.
What analysis settings should I use for Ct (threshold) and baseline with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
We recommend using Auto Ct (Auto threshold) and auto baseline until the Ct cutoff for each target has been set through your clinical validation study.
Can I use the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay to screen for XLA?
We have not validated detection of XLA with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay.
Can I use the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay to determine quantitative results for the KREC or TREC targets?
The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay can be used for semi-quantitative analysis for KREC or TREC targets if you use a standard curve.
Can I use the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay to determine SMA type 0-4 by quantifying SMN2 copy number?
No, the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are not designed to determine SMN2 copy number.
Can I use the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay to determine SMA carrier status?
No, the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are not intended to determine SMA carrier status.
How do you define a normal SMA result in the context of the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
SMA testing determines the absence or presence of SMN1. A person who is considered normal has ≥1 copy of SMN1. Conversely, a person with SMA has a homozygous deletion of SMN1. The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay determine a “Yes/No” result. Presence of SMN1 amplification curves indicates a normal (unaffected) sample, and an absence of SMN1 amplification curves (i.e., undetermined SMN1 Ct) indicates an affected sample.
If my sample has 1 or more copies of SMN2, will the SMN1 target amplification curve for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay be affected?
The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are specific to SMN1. SMN2 does not impact SMN1 amplification. The assay has been challenged with high copy number of SMN2 target and it did not affect amplification.
What is the purpose of the SMN2 blocker in the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
SMA testing determines the absence or presence of SMN1. A person who is considered normal has ≥1 copy of SMN1. Conversely, a person with SMA has a homozygous deletion of SMN1. SMN1 and paralog SMN2 differ by five bases, including a single coding base pair (exon 7 C>T) used to detect the deletion of SMN1 or gene conversion to SMN2. Therefore, specificity of SMN1 is important, as cross hybridization of SMN2 can cause a false positive signal of SMN1 presence. Due to the potential cross hybridization to SMN2 gene targets, we have designed this assay to “block” SMN1 probe cross-hybridization to SMN2 genomic targets, and therefore provide high specificity to SMN1.
Why is SMN1 specificity important for SMA screening? How is this related to the SMN2 blocker?
SMA testing determines the absence or presence of SMN1. A person who is considered normal has ≥1 copy of SMN1. Conversely, a person with SMA has a homozygous deletion of SMN1. SMN1 and paralog SMN2 differ by five bases, including a single coding base pair (exon 7 C>T) used to detect the deletion of SMN1 or gene conversion to SMN2. Therefore, specificity of SMN1 is important, as cross hybridization of SMN2 can cause a false positive signal of SMN1 presence. Due to the potential cross hybridization to SMN2 gene targets, we have designed this assay to “block” SMN1 probe cross-hybridization to SMN2 genomic targets, and therefore provide high specificity to SMN1.
Why is my exported JSON file for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay saving with the wrong file extension?
The exported JSON file should have a .JSON file extension. If the exported JSON file does not have the .JSON extension once it is exported, click export again, but do not change the file name. Save the exported JSON file in its final location and then change the file name. Changing the JSON file name before saving may alter its file extension.
Can I use a JSON file to analyze my TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay results?
If you are using DA Software v2.4.3 or newer, a JSON file can be created for analysis parameters.
What is the benefit of incorporating a JSON file into my Design and Analysis (DA) Software v2.3.4 or newer for analysis of TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay results?
Incorporating a JSON file into DA Software will allow customers to generate reports from test results. Defined parameters for data analysis can be entered into a JSON file. This JSON file can then be applied to each subsequent sample run, thus allowing for quick and consistent reporting of testing results.
How do I obtain Design and Analysis (DA) Software to run the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
The most current version of the DA Software can be downloaded here: https://www.thermofisher.com/us/en/home/global/forms/life-science/quantstudio-6-7-pro-software.html
What version of Design and Analysis (DA) Software should I use for analysis of data generated with the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
We recommend using DA Software v2.4.3 or newer. The most current version of the software can be downloaded here: https://www.thermofisher.com/us/en/home/global/forms/life-science/quantstudio-6-7-pro-software.html
At what temperature should I store the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay are shipped at ambient temperature (https://assets.thermofisher.com/TFS-Assets/LSG/brochures/cms_081489.pdf) and we recommend storing them at -20 degrees C upon receipt.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, can I use DNA extract obtained from a 1.5 mm punch size on a 384-well format?
Yes, DNA extract obtained from a 1.5 mm punch size can be used on a 384-well format.
Which QuantStudio block types is theTaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay protocol compatible with?
The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay protocol is compatible with all 3 QuantStudio block types: 96-well 0.1 mL, 96-well 0.2 mL, and 384-well.
Which qPCR instruments are the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay protocol compatible with?
The TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay have been validated on the QuantStudio 5, QuantStudio 6 Flex, QuantStudio 7 Flex, QuantStudio 12K Flex, and QuantStudio Dx (in RUO mode) instruments. The assays are also compatible with the QuantStudio 6 Pro and QuantStudio 7 Pro instruments but have not been validated on these instruments.
In the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what should I set my method ramp rate to?
For all QuantStudio instruments, we recommend setting the ramp rate to the default speed.
Can I use standard cycling rather than Fast cycling for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
Yes, standard cycling may be used for the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, is a 2-fold serial dilution appropriate when generating a standard curve?
Assuming ˜100% PCR efficiency, a 2-fold, 5-fold, or 10-fold serial dilution can be used to generate a standard curve. We recommend using a 10-fold serial dilution of known concentration of target constructs.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, how many dilution points are recommended when generating a standard curve?
We recommend using 4-6 dilution points. Each dilution run in triplicates should suffice.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, is it okay to prepare a duplex TREC and KREC plasmid mix to generate a standard curve?
Yes, since the assay itself is multiplex, a single mix of TREC and KREC plasmids can be used to generate standard curves for TREC and KREC simultaneously. If using a 20 µL reaction volume, use 4.5 µL of each plasmid for a total volume of 9 µL. Adjust the calculated copies/reaction accordingly.
What are the storage conditions for dried blood spots (DBS) before use?
For short term storage, we recommend storing dried blood spots (DBS) desiccated or non-desiccated at room temperature.
For long term storage, we recommend storing dried blood spots (DBS) at room temperature or at -20 degrees C.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, for how long can I store the extraction material without seeing significant effects on target amplification?
Based on our stability study data, the extracted lysate from 3.2 mm DBS samples is stable for up to 12 days at RT and 4 degrees C.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, can I use regular TE (with 1 mM EDTA) instead of low EDTA TE (0.1 mM EDTA) to prepare the stabilizing buffer?
Yes, as long as the pH is 8, regular TE (with 1 mM EDTA) or low EDTA TE (0.1 mM EDTA) can be used to prepare the stabilizing buffer. However, low EDTA TE is preferred to minimize interference with the Mg2+ present in the qPCR reaction.
Is there a preferred software program to use when analyzing data from the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay?
We recommend using Design and Analysis (DA) Software v2.4.3. or newer for analyzing data from the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay. Please download the latest version of the software from here: https://www.thermofisher.com/us/en/home/global/forms/life-science/quantstudio-6-7-pro-software.html
In the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, can I use 0.1% Tween 20 in the stabilizing buffer instead of 0.05% Tween 20?
Yes, 0.05% Tween 20 can be used in the stabilizing buffer. We have tested 0%, 0.05%, 0.10%, and 0.5% Tween 20 in the stabilizing buffer, and with all these Tween 20 concentrations, target amplification curves were comparable. At 5% Tween 20 in the stabilizing buffer, SMN1 amplification was affected significantly, while RNaseP and TREC amplification curves were not as affected.
In the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, when removing the supernatant after initial wash, what should I set the pipette to?
The pipette should be set to above 150 µL to ensure that all liquid (eg., 157 µL) is removed from the punch. Leaving liquid in the punch will have negative consequences and may leave out impurities/interfering substances that can impact the SMN1 curve profile.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, once the DBS wash buffer is prepared (PBS + Thesit), can it be stored or does it need to be prepared fresh each day?
The DBS wash buffer will not settle out at RT, so it can be stored.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, how should I heat the Thesit reagent?
The Thesit reagent can be warmed up in a water bath until it liquifies.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, how much Tween 20 should I add to TE Buffer to prepare the stabilizing solution?
We recommend adding 0.5 mL Tween 20 to 100 mL TE (0.005X TE volume) or 120 µL Tween to 24 mL TE.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, can punches (1.5 mm or 3.2 mm) be put directly into 384-well plates or do they have to be transferred?
Our protocol is a transfer protocol, so it will always start in a 96-well plate. However, labs can validate their own methods.
Which punch size (1.5 mm or 3.2 mm) is most commonly used in labs?
According to Centers for Disease Control and Prevention (CDC), 1.5 mm punch size is the most popular in the US.
For the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, can I use QuantaBio ExtracTa instead of 0.5% Thesit wash buffer and Extract All Lysis Solution to extract my DNA?
Yes. Assay performance is comparable if QuantaBio ExtracTa is used instead of 0.5% Thesit wash buffer and Extract All Lysis Solution to extract DNA.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if I use the Extract All Stabilizing Solution (as opposed to the prepared TE Stabilizing Buffer containing 0.05% Tween 20)?
If the Stabilizing Solution from the Extract All Kit is used, target amplification curves will appear S-shaped.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if the DBS wash buffer is prepared to contain more than 0.5% Thesit?
We have tested 0%, 0.5%, and 1.0% Thesit concentration in the DBS wash buffer and have not observed significant effects on the target amplification curves. At 5% Thesit concentration in the DBS wash buffer, SMA assay performance was slightly affected, however, TREC and RNaseP targets amplified well.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if the DBS wash buffer is prepared with 0% Thesit?
There is no significant effect on the target amplification curves with 0% Thesit in the DBS wash buffer.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if the stabilizing buffer is prepared to contain more than 0.05% Tween 20?
We have tested 0%, 0.05%, 0.10%, and 0.5% Tween 20 in the stabilizing buffer, and with all these Tween 20 concentrations, target amplification curves were comparable. At 5% Tween 20 in the stabilizing buffer, SMN1 amplification was affected significantly, while RNaseP and TREC amplification curves were not as affected.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if the stabilizing buffer is prepared with 0% Tween 20?
Having no Tween 20 in the stabilizing buffer does not significantly affect the target amplification curves.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what is the effect on the target amplification curves if I leave the plate on the microplate shaker for more than 30 min?
We have tested 30 min, 1 hr, 2 hr, and 3.5 hr wash duration (on shaker), and we did not observe significant effects on the target amplification with prolonged DBS washing.
In the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, if I plan to use only non-desiccated and freshly prepared samples, do I need to do 30 min of DBS washing?
For non-desiccated and freshly prepared samples, 10 min of DBS washing with wash buffer works well, however, customer will need to validate this modified workflow.
In the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, why does the laboratory plate shaker require 1500 rpm speed?
The validated dried blood spots (DBS) wash protocol includes centrifugation at 1500 rpm for 30 min. This protocol is suitable for various DBS sample types: desiccated, non-desiccated, >7 days from preparation, ≥7 days from preparation (freshly prepared). A higher rpm is required for DBS samples that have been desiccated for long term storage to adequately remove DNA from the DBS punch.
With the TaqMan SCID/SMA Assay and TaqMan SCID/SMA Plus Assay, what color should the dried blood spots (DBS) be after extraction?
Extracted dried blood spots (DBS) punches obtained from freshly prepared samples appear nearly white/cream. If the DBS sample was prepared >7 days ago or has been desiccated for a prolonged period of time, the color may appear light red/brown. If the extracted DBS punch remains a dark red/brown color, additional wash time may be required.