CTS™ B-27™ Supplement XenoFree (50X) - FAQs
CTS™ B-27™ Supplement XenoFree (50X) - FAQs
View additional product information for CTS™ B-27™ Supplement XenoFree (50X) - FAQs (A5047501)
5 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Do you offer CTS media that are compatible with CTS B-27 Supplement Xeno-Free?
Yes, examples include CTS Neurobasal Medium (Cat. No. A13712), CTS Neurobasal-A Medium (Cat. No. A13710), and CTS KnockOut DMEM/F12 (Cat. No. A13708).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I request the traceability documentation for CTS B-27 Supplement Xeno-Free?
For instructions on how to request traceability documentation, please visit the Cell Therapy Systems Support webpage (https://www.thermofisher.com/us/en/home/clinical/cell-gene-therapy/cell-therapy/cell-therapy-systems/cell-therapy-systems-support.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What accompanying documentation is available for CTS B-27 Supplement Xeno-Free?
Traceability documentation including FDA Drug Master Files and/or Regulatory Support Files, certificates of analysis, and certificates of origin are available upon request.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What varieties of CTS B-27 Supplement Xeno-Free are available?
CTS B-27 Supplement Xeno-Free is available in three versions:
- CTS B-27 Supplement Xeno-Free (Cat. No. A5047501)
- CTS B-27 Supplement Xeno-Free minus Vitamin A (50X) (Cat. No. A5047601)
- CTS B-27 Supplement Xeno-Free minus Insulin (50X) (Cat. No. A5047701)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.