TaqCheck™ SARS-CoV-2 Control Dilution Buffer
Applied Biosystems™

TaqCheck™ SARS-CoV-2 Control Dilution Buffer

Applied Biosystems TaqCheck SARS-CoV-2 Control Dilution Buffer is required for use of the TaqCheck SARS-CoV-2 Fast PCR Assay. It isRead more
Have Questions?
Catalog NumberQuantity
A504863 x 250 μL
Catalog number A50486
Price (USD)
-
Quantity:
3 x 250 μL
Applied Biosystems TaqCheck SARS-CoV-2 Control Dilution Buffer is required for use of the TaqCheck SARS-CoV-2 Fast PCR Assay. It is used to dilute the RNA control.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™ 5 Real‐Time PCR Instrument, 384-well
Product LineTaqCheck
Quantity3 x 250 μL
Shipping ConditionDry Ice
SpeciesHuman
TargetSevere acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
For Use With (Application)Real Time PCR (qPCR)
PCR Method1-step RT-qPCR
Reaction SpeedFast
Unit SizeEach