CTS™ Viral-Plex™ Complexation Buffer
Gibco™

CTS™ Viral-Plex™ Complexation Buffer

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagentLeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
A5145401500 mL
A51454021 L
A514540310 L
Número do catálogo A5145401
Preço (BRL)
-
Quantity:
500 mL

Gibco CTS Viral-Plex Complexation Buffer is a chemically defined, protein-free, phenol-red-free buffer for complexing plasmid DNA with a transfection reagent for standard transfection applications. It is a part of the CTS AAV-MAX system and is designed to be used with the CTS AAV-MAX Transfection Kit for the transient transfection of CTS Viral Production Cells 2.0.

This buffer is manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. A Drug Master File and Regulatory Support File are available upon request.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS Viral Production Medium, CTS AAV-MAX Transfection Kit, and Viral Production Medium, AGT.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Especificações
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
PackagingBottle
Recommended Storage500 mL bottle. Store at 2°C to 8°C and protect from light.
Shelf Life12 months from date of manufacture
SterilitySterile-filtered
For Use With (Application)Transient transfection for mammalian AAV production
Physical FormLiquid
Quantity500 mL
Unit SizeEach