CTS™ Viral Production Cells 2.0
Gibco™

CTS™ Viral Production Cells 2.0

Gibco™ CTS Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F cell line and are a core component of the CTS AAV-MAX Helper-Free AAV Production System.
Have Questions?
Catalog NumberQuantity
A484001.5 mL
Catalog number A48400
Price (USD)
-
Quantity:
1.5 mL

Gibco™ CTS Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F cell line and are a core component of the CTS AAV-MAX Helper-Free AAV Production System. The cells have been adapted to the chemically defined CTS Viral Production Medium and are maintained in suspension culture in that medium. CTS Viral Production Cells 2.0 can be transfected using the CTS AAV-MAX Transfection Kit for adeno-associated virus (AAV) production.

Features of the CTS Viral Production Cells 2.0 include:

  • Clonal, 293F-derived, high-producer cell line
  • Absence of SV40 large T antigen
  • Optimized for high-density, suspension culture (>12 million cells/mL) in chemically defined medium
  • Enable production of high AAV titers, greater than 5 x1010 vg/mL (unpurified)
  • Robust scalability and passage stability

CTS Viral Production Cells 2.0 are a fully documented cGMP cell line

  • Fully documented, cGMP-banked with detailed cell line lineage history
  • Extensive quality testing based on USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations
  • Flexible licensing options without the burden of downstream royalties
  • Contact us (outlicensing@thermofisher.com) for more information and access
  • Frozen cells are supplied in a vial containing 1.5 mL of cells at 1 x 107 viable cells/mL in CTS Viral Production Medium and 7.5% DMSO. Cells should be thawed directly into CTS Viral Production Medium for optimal performance.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Medium (A5144001), CTS AAV-MAX Transfection Kit (A5427701), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatCryopreserved vial
Manufacturing Quality21 CFR Parts 210 and 211
Media RecommendationGibco™ CTS™ Viral Production Medium
No. of CellsSee Certificate of Analysis
Product TypeViral Production Cell Line
Quantity1.5 mL
Shelf Life10 years from date of manufacture
Shipping ConditionDry ice or vapor phase liquid nitrogen
Tested ForAAV Production
Cell LineViral Production Cells 2.0
Cell TypeKidney (Embryonic)
Culture TypeSuspension Cell Culture
Unit SizeEach
Contents & Storage
1 x 107 cells in a 2-mL vial. Store in vapor phase liquid nitrogen.

Frequently asked questions (FAQs)

Can I use CTS Viral Production Cells (Cat. No. A53956) with CTS AAV MAX products?

CTS Viral Production Cells 2.0 are specifically designed to work with CTS AAV MAX products to provide maximal adeno-associated virus (AAV) titers and optimal performance. CTS Viral Production Cells can be used but we don't recommend doing this as it will reduce system performance.

What is the highest density that CTS Viral Production Cells 2.0 can be cultured at?

CTS Viral Production Cells 2.0 achieve maximum cell densities of >1.2 x 10E7 cells/mL in shake flask cultures.

What is the passage stability of CTS Viral Production Cells 2.0?

We recommend using CTS Viral Production Cells 2.0 between passages 3 to 25 for adeno-associated virus (AAV) production.

How long do CTS Viral Production Cells 2.0 require to recover post-thaw before transfection?

We recommend allowing freshly thawed CTS Viral Production Cells 2.0 to recover in culture for at least three passages prior to transfection.

Are CTS Viral Production Cells 2.0 a clonal cell line?

Yes, CTS Viral Production Cells 2.0 cells are a clonal cell line derived from single cell cloning of 293-F cells.