DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX), 50 mg - FAQs
DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX), 50 mg - FAQs
View additional product information for Mass Spectrometry Compatible Crosslinkers - FAQs (A52287, A52286)
2 product FAQs found
What is the advantage of using phospho-enrichable crosslinkers?
Phospho-enrichable crosslinkers such as DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) and TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX) are great tools for studying protein-protein Interaction, protein 3D structure, and for mass spectrometry (MS) analysis. They can be used to enrich low-abundance crosslinked peptides and thereby improve MS identification rates.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) cell-permeable?
DSPP is not cell-permeable. We offer TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX), Cat. No. A52287 that is a cell-permeable phopho-enrichable crosslinker.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.