AccelerOme™ System Test Kit - FAQs
AccelerOme™ System Test Kit - FAQs
View additional product information for AccelerOme™ System Test Kit - FAQs (A53220)
4 product FAQs found
Can the AccelerOme System Test Kit be used on a different platform such as Agilent?
The AccelerOme System Test Kit has been optimized exclusively for the AccelerOme Automated Sample Prep Platform.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How often do I need to use the AccelerOme System Test Kit?
The AccelerOme System Test Kit is meant to be used for calibration (by FAS/FSE) when the AccelerOme Automated Sample Prep System is installed or restarted after maintenance.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I reuse the AccelerOme System Test Kit?
We do not recommend reusing the AccelerOme System Test Kit. It is meant to be used for calibration (by FAS/FSE) when the AccelerOme Automated Sample Prep System is installed or restarted after maintenance.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
In which format is the AccelerOme System Test Kit provided?
The AccelerOme System Test Kit is a ready-to-use tray to be inserted in the AccelerOme Automated Sample Prep System, to start the calibration.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.