Oncomine™ Dx Express Test Panel
Ion Torrent™

Oncomine™ Dx Express Test Panel

The Oncomine Dx Express Test Panel contains a set of oligos intended for use with the required instruments and consumables.
Have Questions?
Catalog NumberNo. of Reactions
A5410332 Reactions
Catalog number A54103
Price (MXN)
54,361.70
Each
No. of Reactions:
32 Reactions
Price (MXN)
54,361.70
Each

The Oncomine Dx Express Test is an in vitro diagnostic assay that uses targeted next-generation sequencing (NGS) to detect biomarkers across 46 genes. The Oncomine™ Dx Express Test is a qualitative in vitro diagnostic test that uses targeted next generation sequencing (NGS) technology to detect substitutions, insertions, and deletions in 42 genes from DNA, copy number variants (CNVs) in 10 genes from DNA, and fusions or splice variants in 18 genes from RNA isolated from formalin-fixed, paraffin-embedded (FFPE) tumor samples using the Genexus™ Dx Integrated Sequencer.

The test is designed for use with the Genexus Dx System. Its fully automated workflow—from sample preparation to results—can be completed in as little 24 hours, supporting timely decision-making in clinical practice.

Additional products that are required to complete the Oncomine Dx Express Test workflow include:

  • Genexus Dx Integrated Sequencer
  • Genexus Dx Library Strips 1 and 2-HD
  • Genexus Dx Templating Strips 3-GX5 and 4
  • Genexus Dx Sequencing Kit
  • Genexus Dx Barcodes 1-32 HD
  • Genexus Dx GX5 Chip and Genexus Coupler
  • Genexus Dx Pipette Tips
  • Oncomine Express Test FFPE DNA and RNA Control Kit
  • Oncomine Express Test Plasma cfTNA Control Kit*
  • MicroAmp EnduraPlate Optical 96-Well Clear GPLE Reaction Plates with Barcode
  • Thermo Scientific Adhesive PCR Plate Foils

*Not available in the U.S.

For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
Chip CompatibilityGenexus GX5 Dx
For Use With (Application)Oncology Diagnostics
For Use With (Equipment)Genexus Dx Integrated Sequencer
LibrariesAmplicon
No. of Reactions32 Reactions
No. of SamplesUp to 14 FFPE or 20 Plasma Samples
Nucleic Acid TypeDNA, RNA, or cfTNA
Product LineOncomine
Product TypeAssay
Quantity1 Ea.
Sample TypeFFPE Tumor Tissue, Plasma*
ThroughputUp to 14 FFPE or 5 Plasma Samples per GX5 Chip
Workflow StepPanel
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

How many genes and variant types are covered by the Oncomine Dx Express Test?

The Oncomine Dx Express Test (Cat. No. A54103) detects biomarkers across 46 unique genes. It detects substitutions, insertions, and deletions in 42 genes from DNA; copy number variants in 10 genes from DNA; and fusions or splice variants in 18 genes from RNA isolated from FFPE tumor samples.

Which PTEN gene alterations can the Oncomine Dx Express Test detect?

The Oncomine Dx Express Test (Cat. No. A54103) detects predefined PTEN substitutions, insertions, and deletions from DNA isolated from FFPE tumor samples. Refer to the Oncomine Dx Express Test User Guide for the complete list of validated PTEN variants.

How many DNA and RNA FFPE samples can I run with the Oncomine Dx Express Test?

Up to 14 FFPE DNA and RNA samples can be included in a sequencing run. The run also includes positive and no-template controls (NTCs) for both the DNA and RNA assays.

Which genes are included in the Oncomine Dx Express Test Panel?

The Oncomine Dx Express Test Panel is a targeted next-generation sequencing (NGS) assay that evaluates 46 genes across DNA and RNA. It detects multiple classes of genomic alterations, including single nucleotide variants (SNVs), insertions/deletions (indels), copy number variants (CNVs), gene fusions, and splice variants.

For the Oncomine Dx Express Test Panel, do the 32 reactions mean 16 DNA and 16 RNA reactions?

The 32 reactions can support up to 14 FFPE (DNA + RNA) samples or up to 20 plasma samples in the CE IVD version of ODxET. The FDA approved version does not support liquid biopsy and can be used for 14 FFPE samples. Some of the reactions are used for controls.