TrueMark™ STI Select Panel Combo Kit - FAQs
View additional product information for TrueMark™ STI Select Panel Combo Kit - FAQs (A57083)
5 product FAQs found
Can I order the assays in the TrueMark STI Select Panel Combo Kit (Cat. No. A57083) separately?
No, the assay IDs for the individual target assays in the TrueMark STI Select Panel are not provided separately.
Do I need to use RNase P with the assays included in the TrueMark, STI Select Panel, Combo Kit?
Yes, RNase P is the human internal sample control target utilized in the TrueMark, STI Select Panel, Combo Kit and it is one of the multiplexed assays in the kit.
Which instruments, consumables, and reagents do I need for the TrueMark STI Select Panel, Combo Kit? Where can I find the full list of products and protocols?
We recommend running the TrueMark STI Select Panel, Combo Kit on the QuantStudio 5 Real-Time PCR instrument, 96-well, 0.2 mL block though other instruments that have the ability to detect Alexa Fluor 647 dye are capable of running the kit. Information about other reagents and consumables that are needed can be found in the user guide ((https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).
Do you provide run templates for the TrueMark STI Select Panel, Combo Kit?
No, these are single-tube assays and the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf) provides details on how to setup and create a template to detect and assign the targets to each well.
What are the components of the TrueMark STI Select Panel, Combo Kit? Are there any other required materials that are not supplied?
The TrueMark STI Select Panel, Combo Kit (Cat. No. A57083) contains the TrueMark STI Select Panel (Part No. A52691) and the TrueMark Infectious Disease 1-Step Multiplex Master Mix (No ROX) (Part No. A57052). Other required materials are standard qPCR consumables and instruments and they can be found in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).