Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          1. Home▸
          2. Shop All Products▸
          3. PCR Equipment and Supplies▸
          4. PCR Supplies▸
          5. Real Time PCR Products▸
          6. Real Time PCR Reagents and Kits▸
          7. TrueMark™ STI Select Panel Combo Kit
          Request Quote
          Share
          TrueMark™ STI Select Panel Combo Kit
          Applied Biosystems™

          TrueMark™ STI Select Panel Combo Kit

          The Applied Biosystems TrueMark STI Select Panel Combo Kit is a multiplex real‐time PCR solution that contains the assay and master mix needed for the simultaneous detection of RNA in extracted human samples.
          Have Questions?
          Catalog NumberQuantity
          A57083200 reactions
          Catalog number A57083
          Price (USD)
          4,105.00
          Each
          Quantity:
          200 reactions
          Price (USD)
          4,105.00
          Each
          The Applied Biosystems TrueMark STI Select Panel Combo Kit is a multiplex real‐time PCR solution that contains the assay and master mix needed for the simultaneous detection of nucleic acid from Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG), Mycoplasma genitalium (MG), and Trichomonas vaginalis (TV) in extracted human samples. Human ribonuclease P (RNase P) is included as endogenous sample collection control.

          Features of the TrueMark STI Select Panel Combo Kit include:
          • Single test for CT, NG, MG, TV; detect and differentiate between clinically relevant targets found in sexually transmitted infections
          • Affordable and scalable—add to existing infrastructure to expand menu while maintaining low operational costs
          • Ready-to-use off-the-shelf solution—seamlessly integrate test into the existing workflow
          • Research Use Only

          The TrueMark STI Select Panel Combo Kit includes the TrueMark STI Select Panel and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark STI Amplification Control, the kit can be used to quickly evaluate up to 200 samples and is validated for use with RNA extracted from vaginal swabs. The kit is recommended for use with the MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit, the KingFisher Flex Purification System, and the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block).

          The TrueMark STI Select Panel Combo Kit includes the following components:
          • TrueMark STI Select Panel—multiplexed assay that contains primer and probe sets specific to CT, NG, MG, TV, and RNase P targets
          • TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)

          Use this kit with:
          • TrueMark STI Amplification Control (Cat. No. A57009)—plasmid control containing templates specific to CT, NG, MG, TV, and RNase P

          Specifications
          For Use With (Equipment)QuantStudio™ 5 Real‐Time PCR System, 96-well, 0.2-mL block
          No. of Reactions200 Reactions
          Quantity200 reactions
          Detection MethodPrimer-probe
          For Use With (Application)Pathogen Detection
          Unit SizeEach

          Frequently asked questions (FAQs)

          Can I order the assays in the TrueMark STI Select Panel Combo Kit (Cat. No. A57083) separately?

          No, the assay IDs for the individual target assays in the TrueMark STI Select Panel are not provided separately.

          Do I need to use RNase P with the assays included in the TrueMark, STI Select Panel, Combo Kit?

          Yes, RNase P is the human internal sample control target utilized in the TrueMark, STI Select Panel, Combo Kit and it is one of the multiplexed assays in the kit.

          Which instruments, consumables, and reagents do I need for the TrueMark STI Select Panel, Combo Kit? Where can I find the full list of products and protocols?

          We recommend running the TrueMark STI Select Panel, Combo Kit on the QuantStudio 5 Real-Time PCR instrument, 96-well, 0.2 mL block though other instruments that have the ability to detect Alexa Fluor 647 dye are capable of running the kit. Information about other reagents and consumables that are needed can be found in the user guide ((https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).

          Do you provide run templates for the TrueMark STI Select Panel, Combo Kit?

          No, these are single-tube assays and the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf) provides details on how to setup and create a template to detect and assign the targets to each well.

          What are the components of the TrueMark STI Select Panel, Combo Kit? Are there any other required materials that are not supplied?

          The TrueMark STI Select Panel, Combo Kit (Cat. No. A57083) contains the TrueMark STI Select Panel (Part No. A52691) and the TrueMark Infectious Disease 1-Step Multiplex Master Mix (No ROX) (Part No. A57052). Other required materials are standard qPCR consumables and instruments and they can be found in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf).

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States