Product Image
Applied Biosystems™
MicroSEQ™ ID 16S rDNA 500 Library v2022
The MicroSEQ ID 16S rDNA 500 Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0.
Have Questions?
카탈로그 번호수량
A573121 Ea.
카탈로그 번호 A57312
제품 가격(KRW)
-
수량:
1 Ea.
The MicroSEQ ID 16S rDNA 500 Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 2100 reference 16S rRNA sequences from bacteria. The library entries contain the first 500 bases of the 16S rRNA gene, enough to identify bacterial unknowns.
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)Microbial Identification
용도(장비)SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
라이선스Static
운영 체제Microsoft Windows 10
제품라인MicroSeq™ ID
제품 유형Microbial Identification Software
수량1 Ea.
소프트웨어Data Collection and Analysis Software
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
자주 묻는 질문(FAQ)
What are the operating system requirements for MicroSEQ ID 16S rDNA 500 Library v2022?
The operating system requirements for MicroSEQ ID 16S rDNA 500 Library v2022 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicroSEQ ID 16S rDNA 500 Library v2018 and v2022?
MicroSEQ ID 16S rDNA 500 Library v2022 contains identical content to the previous generation library, MicroSEQ ID 16S rDNA 500 Library v2018. MicroSEQ ID 16s rDNA 500 Library v2022 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID 16s rDNA 500 Library v2018 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the browser compatibility requirement for MicroSEQ ID 16S rDNA 500 Library v2022?
MicroSEQ ID 16S rDNA 500 Library v2022 is compatible with Google Chrome v108.0.5359.73.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Windows OS (Operating System) is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Microbial Identification Software v4.0?
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.