What are the operating system requirements for MicroSEQ ID 16S rDNA 500 Library v2022?
The operating system requirements for MicroSEQ ID 16S rDNA 500 Library v2022 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.