What is the difference between MicroSEQ ID Fungal Gene Library v2018 and v2022?
MicroSEQ ID Fungal Gene Library v2022 contains identical content to the previous generation library, MicroSEQ ID Fungal Gene Library v2018. MicroSEQ ID Fungal Gene Library v2022 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID Fungal Gene Library v2018 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.