Product Image
Applied Biosystems™
MicroSEQ™ ID Fungal Gene Library v2022
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0.
Have Questions?
Catalog NumberQuantity
A573141 Ea.
Catalog number A57314
Price (MXN)
-
Quantity:
1 Ea.
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microbial Identification
For Use With (Equipment)SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
LicenseStatic
Operating SystemMicrosoft Windows 10
Product LineMicroSeq™ ID
Product TypeMicrobial Identification Software
Quantity1 Ea.
SoftwareData Collection and Analysis Software
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Frequently asked questions (FAQs)
What is the difference between MicroSEQ ID Fungal Gene Library v2018 and v2022?
MicroSEQ ID Fungal Gene Library v2022 contains identical content to the previous generation library, MicroSEQ ID Fungal Gene Library v2018. MicroSEQ ID Fungal Gene Library v2022 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID Fungal Gene Library v2018 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the browser compatibility requirement for MicroSEQ ID Fungal Gene Library v2022?
MicroSEQ ID Fungal Gene Library v20222 is compatible with Google Chrome v108.0.5359.73.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the operating system requirements for MicroSEQ ID Fungal Gene Library v2022?
The operating system requirements for MicroSEQ ID Fungal Gene Library v2022 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Windows OS (Operating System) is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Microbial Identification Software v4.0?
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.