Culturing spheroids in bioreactors is similar to culturing spheroids in small-scale vessels. The seeding density can remain the same (200,000 cells/mL), along with the inclusion of Y-27632 and DNase I. By following these recommendations, the spheroids can be grown for approximately 5 days. For additional considerations, follow the recommendations below for different types of bioreactors:
Vertical wheel bioreactor:
Working volume: 2-3 L
RPM: 20-25 RPM
Spheroid sedimentation time: Up to 10 mins
CTS TrypLE Select Enzyme concentration: 0.5X-0.75X
CTS TrypLE Select Enzyme dissociation volume: 50-60 mL
CTS TrypLE Select Enzyme dissociation time: 30-40 mins
Horizontal blade impeller bioreactor:
Working volume: 2-3 L
RPM: 65 RPM (first day); 130 RPM final (ramp over next day)
Spheroid sedimentation time: Up to 10 mins
CTS TrypLE Select Enzyme concentration: 0.5X-0.75X
CTS TrypLE Select Enzyme dissociation volume: 50-60 mL
CTS TrypLE Select Enzyme dissociation time: 30-40 mins
Find additional tips, troubleshooting help, and resources within our
Cell Culture Support Center.