Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
CTS™ StemScale™ PSC Suspension Medium
Gibco™

CTS™ StemScale™ PSC Suspension Medium

CTS (Cell Therapy Systems) StemScale PSC Suspension Medium is the first commercially available medium specifically developed for scalable suspension cultureRead more
Have Questions?
Catalog NumberQuantity
A58696011 L
Catalog number A5869601
Price (USD)
1,732.00
Each
Quantity:
1 L
Price (USD)
1,732.00
Each

CTS (Cell Therapy Systems) StemScale PSC Suspension Medium is the first commercially available medium specifically developed for scalable suspension culture of pluripotent stem cells (PSCs) for clinical manufacturing and commercialization of PSC-derived therapies. With an optimized and easy-to-use protocol, coupled with a xeno-free formulation, CTS StemScale medium provides a fast, easy, and cost-effective solution for scaling from translational studies to clinical manufacturing.  PSCs expanded in CTS StemScale medium maintain pluripotency and genomic stability across multiple passages and can be differentiated to the three germ layers. 

CTS StemScale PSC Suspension Medium provides:

  • Maximal expansion capability versus other PSC suspension media, helping reduce manufacturing time and cost
  • Scalable expansion of PSCs for clinical manufacturing and automation platforms
  • Improved cell viability and yield with an optimized protocol
  • Cell therapy manufacturing with extensive product safety and quality testing, GMP manufacturing, traceability of raw materials, and regulatory documentation
  • PSC suspension culture intellectual property (IP)

Maximal expansion capability
PSCs cultured in CTS StemScale medium can achieve 5–10-fold expansion per passage and up to 30% higher expansion versus other PSC suspension media.  Long-term expansion of PSCs in CTS StemScale medium (>30 passages) can be accomplished without the loss of expansion potential, pluripotency, or genomic stability. 

Scalable expansion
CTS StemScale medium promotes consistent growth of PSCs in plates, flasks, and bioreactors.  PSCs can be efficiently expanded in CTS StemScale medium across vessel types using the same culture parameters at both small and large scales.

Optimized protocol
CTS StemScale medium promotes spheroid nucleation of both induced pluripotent stem cells (iPSCs) and embryonic stem cells (ESCs).  PSCs grown in CTS StemScale medium are fed by a 50% medium replacement strategy. This method reduces metabolic waste build-up while maintaining consistent media volumes in the culture, a critical factor in the maintenance of spheroid size.  During passaging, the use of cell strainers is not required when dissociating the PSC spheroids, thereby helping minimize cell loss and improving cell viability while supporting maximal expansion of PSCs.

GMP manufactured
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available for CTS StemScale PSC Suspension Medium.

PSC suspension culture IP
CTS StemScale is the only commercially available PSC culture medium that provides customers rights to Thermo Fisher’s exclusively licensed foundational intellectual property for culturing and expanding PSCs in suspension.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeEmbryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Concentration1X (CTS StemScale Basal Medium), 10X (CTS StemScale Supplement)
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeCell Culture Medium
Quantity1 L
Shelf Life12 Months from Date of Manufacture
Shipping ConditionAmbient (CTS StemScale Basal Medium), Dry Ice (CTS StemScale Supplement)
ClassificationXeno-free
Culture TypeSuspension Cell Culture, Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
• CTS StemScale Basal Medium, 900 mL; Store at 2°C to 8°C and protect from light.
• CTS StemScale Supplement, 100 mL; Store at -5°C to -20°C and protect from light.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

How does the CTS StemScale PSC Suspension Medium passaging protocol compare to protocols for other suspension culture media?

The CTS StemScale PSC Suspension Medium protocol recommends passaging every 5 - 6 days, depending on spheroid size. We recommend passaging suspension cultures when the average spheroid diameter is between 300 - 400 µm, which occurs approximately after 5 – 6 days of growth. Passaging earlier is also an option, although the final cell yield will be lower than what is typically obtained.

Passaging protocols for other suspension culture media may offer less flexibility, depending upon whether they utilize a fed-batch or overlay strategy. Generally, these cultures require a strict passaging schedule with little room for flexibility on the weekend.

The CTS StemScale PSC Suspension Medium passaging protocol also does not require the use of cell strainers, while some other PSC suspension culture media protocols may specifically require these devices.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are there advantages to the CTS StemScale PSC Suspension Medium feeding method as compared to those for other media for suspension cultures?

The CTS StemScale PSC Suspension Medium feeding method involves replacing 50% of the spent medium with fresh medium. This feeding method prevents spheroids from being cultured in medium that is accumulating significant quantities of waste by-products.

Feeding methods which use fed-batch or overlay strategy do not remove the spent medium from suspension cultures. These methods reduce hands-on time when feeding cultures; however, the accumulation of waste by-products is likely to negatively impact the health of the cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Managing multiple components in a medium kit can be challenging. Does the CTS StemScale PSC Suspension Medium kit have a similar number of components as others?

The CTS StemScale PSC Suspension Medium kit consists of only two components: a basal medium and a supplement. Following reconstitution, the complete medium is used for all steps, from initiating cultures to feeding cultures.

Other suspension culture medium kits sometimes consist of multiple components, which may be utilized at different points in the suspension culture process.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What would the CTS StemScale PSC Suspension Medium cultures look like if I use diluted CTS TrypLE Select Enzyme without including DNase I?

Some cells may lyse and release genomic DNA into the suspension culture environment during dissociation with CTS TrypLE Select Enzyme. Without DNase I, the presence of small quantities of genomic DNA will cause some spheroids to aggregate into a single clump.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.