Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NF1 Antibodies

          Bethyl Laboratories

          Neurofibromin 1 Recombinant Rabbit Monoclonal Antibody (BLR345M)

          View all (21) NF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Neurofibromin 1 Recombinant Rabbit Monoclonal Antibody (BLR345M)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          Neurofibromin 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Neurofibromin 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neurofibromin 1 Antibody (A700-345) in WB

          Detection of human Neurofibromin-1 by western blot. Samples: Whole cell lysate (10 µg) from LNCaP, U2OS, HT-29, K-562, and HEK293T cells prepared using NETN lysis buffer. Antibody: Rabbit anti-Neurofibromin-1 recombinant monoclonal antibody [BLR345M] (A700-345) used at 1:1,000. Secondary: HRP-conjugated goat anti-rabbit IgG (A120-101P). Detection: Chemiluminescence with an exposure time of 3 seconds. Lower Panel: Rabbit anti-Actin recombinant monoclonal antibody [BLR057F] (A700-057). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neurofibromin 1 Antibody in Western Blot (WB)
          Neurofibromin 1 Antibody in Western Blot (WB)
          Neurofibromin 1 Antibody in Western Blot (WB)
          Neurofibromin 1 Antibody in Flow Cytometry (Flow)
          Neurofibromin 1 Antibody in Flow Cytometry (Flow)
          Neurofibromin 1 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Neurofibromin 1 Recombinant Rabbit Monoclonal Antibody (BLR345M)

          Product Details

          A700-345

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Flow Cytometry (Flow)

          1 µL/1x10^6 cells
          -

          Immunoprecipitation (IP)

          20 µL/mg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          BLR345M

          Immunogen

          Recombinant antibody was purified from cell culture supernatant. Immunogen was a peptide representing a region between residue 2789 and the C-terminus (residue 2839) of human Neurofibromin 1 using the numbering given in entry NP_001035957.1 (Gene ID 4763).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          100 µg/mL

          Amount

          10 µg

          Purification

          purified

          Storage buffer

          PBS, pH 6.8-7.4, with 0.1% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp686J1293; FLJ21220; neurofibromatosis 1; neurofibromatosis-related protein NF-1; unnamed protein product

          View more View less

          Gene Aliases: Dsk9; E030030H24Rik; Mhdadsk9; Nf-1; NFNS; VRNF; WSS

          View more View less

          UniProt ID: (Human) P21359, (Mouse) Q04690

          View more View less

          Entrez Gene ID: (Human) 4763, (Mouse) 18015

          View more View less

          Function(s)
          GTPase activator activity protein binding phosphatidylethanolamine binding phosphatidylcholine binding microtubule binding syndecan binding
          Process(es)
          MAPK cascade osteoblast differentiation metanephros development response to hypoxia liver development negative regulation of endothelial cell proliferation regulation of cell-matrix adhesion cell communication Ras protein signal transduction negative regulation of neuroblast proliferation brain development peripheral nervous system development heart development visual learning Schwann cell development spinal cord development forebrain astrocyte development cerebral cortex development myelination in peripheral nervous system actin cytoskeleton organization extracellular matrix organization collagen fibril organization adrenal gland development negative regulation of cell migration wound healing positive regulation of apoptotic process negative regulation of MAPK cascade pigmentation protein kinase B signaling positive regulation of neuron apoptotic process regulation of blood vessel endothelial cell migration positive regulation of GTPase activity regulation of bone resorption regulation of glial cell differentiation regulation of angiogenesis negative regulation of fibroblast proliferation sympathetic nervous system development camera-type eye morphogenesis negative regulation of oligodendrocyte differentiation smooth muscle tissue development artery morphogenesis forebrain morphogenesis regulation of developmental process cognition regulation of intracellular signal transduction positive regulation of endothelial cell proliferation negative regulation of cell-matrix adhesion negative regulation of protein kinase activity skeletal muscle tissue development negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors regulation of gene expression positive regulation of neuron projection development Schwann cell differentiation negative regulation of angiogenesis amygdala development neural tube development regulation of synaptic transmission, GABAergic cellular response to heat negative regulation of Rac protein signal transduction regulation of cell proliferation negative regulation of protein import into nucleus negative regulation of MAP kinase activity regulation of MAPK cascade regulation of neuron differentiation negative regulation of osteoclast differentiation positive regulation of adenylate cyclase activity negative regulation of Ras protein signal transduction negative regulation of neurotransmitter secretion regulation of long-term neuronal synaptic plasticity negative regulation of astrocyte differentiation hair follicle maturation regulation of synapse organization long-term synaptic potentiation gamma-aminobutyric acid secretion, neurotransmission glutamate secretion, neurotransmission regulation of ERK1 and ERK2 cascade observational learning postsynaptic signal transduction regulation of modification of postsynaptic structure regulation of postsynapse organization regulation of long-term synaptic potentiation positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0