Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
A more sustainable equivalent is available.
Low Profile Tubes, strips of 8
Thermo Scientific™

Low Profile Tubes, strips of 8

Thermo Scientific Strips of 8 Low Profile Tubes reduces the 'dead space' between the heated lid of the thermal cyclerRead more
Have Questions?
Catalog NumberQuantity
AB0771120 strips
Catalog number AB0771
Price (USD)
179.00
Each
Quantity:
120 strips
Request bulk or custom format
Price (USD)
179.00
Each
Thermo Scientific Strips of 8 Low Profile Tubes reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL. Contents: 10 packs of 12 strips.

Features of Thermo Scientific Strips of 8 Low Profile Tubes

• Suitable for 0.2 mL thermal cycler blocks
• 150 μL maximum tube capacity when capped
• Low profile to reduce dead space and increase PCR efficiency
• Labelled A–H end tabs (8-strip)
• Sealable with Flat, Domed, and Ultra Clear Cap Strips

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Low Profile Tubes and Domed Caps, strips of 8
Low Profile Tubes and Domed Caps, strips of 8, assorted colors
Low Profile Tubes and Flat Caps, strips of 8
Low Profile Tubes and Flat Caps, strips of 8, assorted colors
Low Profile Tubes and Ultra Clear Caps, strips of 8
Low Profile Tubes (white) and Ultra Clear Caps, strips of 8
Low Profile Tubes and Domed Caps, strips of 12
Low Profile Tubes and Flat Caps, strips of 12
Low Profile Tubes, strips of 8, white
Low Profile Tubes, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format120 Strip Tubes
Quantity120 strips
Reaction SpeedLow profile
Volume (Metric) Working0.1 mL
Closure TypeNot Included
ColorNatural
MaterialPolypropylene
Product Type8-Strip
SterilityNon-sterile
Volume (Metric)0.15 mL
Unit SizeEach