Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Tubes (white) and Ultra Clear Caps, strips of 8
Thermo Scientific™

Tubes (white) and Ultra Clear Caps, strips of 8

Thermo Scientific Tubes white and Ultra Clear Caps, Strips of 8 are designed for PCR and qPCR applications, and areRead more
Have Questions?
Catalog NumberQuantity
AB119110 Packs of 12 Strips
Catalog number AB1191
Price (USD)
129.65
Online exclusive
224.00
Save 94.35
Each
Quantity:
10 Packs of 12 Strips
Price (USD)
129.65
Online exclusive
224.00
Save 94.35
Each
Thermo Scientific Tubes white and Ultra Clear Caps, Strips of 8 are designed for PCR and qPCR applications, and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers. The 8-strip tubes can be divided into individual tubes.

Features of Thermo Scientific Tubes white and Ultra Clear Caps strips:

• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when capped
• Ideal for use in 96-well V-bottom thermal cyclers
• 10 packs of 12

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Tubes and Domed Caps, strips of 8
Tubes and Domed Caps, strips of 8, blue
Tubes and Domed Caps, strips of 8, green
Tubes and Domed Caps, strips of 8, red
Tubes and Domed Caps, strips of 8, yellow
Tubes and Flat Caps, strips of 8
Tubes and Flat Caps, strips of 8, assorted colors
Tubes and Ultra Clear Caps, strips of 8
Tubes and Domed Caps, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers or Real-Time PCR Systems
Format120 Strip Tubes and Caps
No. of Wells96
Quantity10 Packs of 12 Strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeFlat Cap
ColorWhite, Ultra Clear
MaterialPolypropylene
Product Type8-strip Tube
Volume (Metric)0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

Why should I use Thermo Scientific white plastics in qPCR?

White plates and tubes reflect significantly more signal back to the qPCR instrument's detector than traditional clear plates. The improved signal reflection ensures that even the lowest levels of fluorescence are detected. White well walls also prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed. This keeps variations in the cycler block from affecting qPCR data. Using Thermo Scientific white PCR plastics increases sensitivity and reduces variability within the qPCR assay.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.