Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Aggrecan Antibodies

          Invitrogen

          Aggrecan Monoclonal Antibody (969D4D11)

          9 Published Figures
          43 References
          View all (20) Aggrecan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Aggrecan Monoclonal Antibody (969D4D11)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (9)
          Aggrecan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Aggrecan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Aggrecan Antibody (AHP0022) in ICC/IF

          Regenerative biomaterials 2020 - Figure 1 Immunophenotypic and morphological features of MSCs and chondrocytes in direct co-culture. (A) Immunohistochemical staining of type I, II, X collagen and aggrecan in MSCs/chondrocytes after 17 days of differentiation in direct 2D and 3D co-culture model (EnVision technique, scale bar = 50 um); (B) Chondrocytes and (G) direct co-culture on culture flask (inverted phase contrast microscope, scale bar... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Aggrecan Antibody in Immunocytochemistry (ICC/IF)
          Aggrecan Antibody in Immunocytochemistry (ICC/IF)
          Aggrecan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aggrecan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Aggrecan Antibody in Immunohistochemistry (IHC)
          Aggrecan Antibody in Immunohistochemistry (IHC)
          Aggrecan Antibody in Immunohistochemistry (IHC)
          Aggrecan Antibody in Western Blot (WB)
          Aggrecan Antibody in Western Blot, ELISA (WB, ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Aggrecan Monoclonal Antibody (969D4D11)

          Product Details

          AHP0022

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 5 publications 5 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          View 9 publications 9 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Mass Spectrometry (MS)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 6 publications 6 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Bovine, Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          969D4D11

          Immunogen

          Human natural aggrecan (proteoglycan), purified from human articular cartilage.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          500 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.5mg/mL sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          RRID

          AB_2536346

          Product Specific Information

          This product is recommended as a detection antibody in Sandwich ELISA applications.

          This clone can also be referred to as clone number 969D 4D11 2A9.

          Target Information

          Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cartilage-specific proteoglycan core protein; chondroitin sulfate proteoglycan core protein 1; large aggregating proteoglycan; proteoglycan core protein; unnamed protein product

          View more View less

          Gene Aliases: AGC1; AGCAN; CSPG1; CSPGCP; MSK16; SEDK; SSOAOD

          View more View less

          UniProt ID: (Human) P16112

          View more View less

          Entrez Gene ID: (Human) 176

          View more View less

          Function(s)
          extracellular matrix structural constituent protein binding hyaluronic acid binding extracellular matrix structural constituent conferring compression resistance
          Process(es)
          skeletal system development proteolysis cell adhesion central nervous system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch