MagMAX™ Total Nucleic Acid Isolation Kit - FAQs
MagMAX™ Total Nucleic Acid Isolation Kit - FAQs
View additional product information for MagMAX™ Total Nucleic Acid Isolation Kit - FAQs (AM1840)
7 product FAQs found
Are MagMax kits compatible with RNAlater reagent-treated samples?
Only MagMax96 for Microarrays Total RNA Isolation Kit (Cat. No. AM1839) is compatible, since it uses Trizol reagent and most of the salt carry-over from RNAlater reagent is eliminated during the lysis steps and partitioned from the aqueous RNA layer on top. For other MagMax kits, optimization is needed. High salt carry over is not optimal for MagMax RNA isolation steps.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
Do you offer the components of the MagMAX Total Nucleic Acid Isolation Kit (Cat. No. AM1840) as standalone products?
The following components from the MagMAX Total Nucleic Acid Isolaton Kit can be purchased as standalone products and here are the details:
- NA Binding Beads: Can be purchased separately as DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit, Cat. No. 4489112 (note that these beads are the same as the beads in Cat. No. AM1840, but the fill volume differs)
- Lysis/binding Solution Concentrate: Can be purchased separately as MagMAX Lysis/Binding Solution Concentrate, Cat. No. AM8500
- Wash Solution 1 Concentrate: Can be purchased separately as MagMAX Wash Solution 1 Concentrate, Cat. No. AM8504
- Wash Solution 2 Concentrate: Can be purchased separately as MagMAX Wash Solution 2 Concentrate, Cat. No. AM8640
Can I edit the manufactured KingFisher Flex protocols?
No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.
Can I use magnetic beads from other vendors with the KingFisher Flex instrument?
For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?
Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Can I scale up my reaction volume with the KingFisher Flex instrument?
Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.
Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?
Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.