Mouse RiboPure™-Blood RNA Isolation Kit, 25 preps - FAQs
Mouse RiboPure™-Blood RNA Isolation Kit, 25 preps - FAQs
View additional product information for Mouse RiboPure™-Blood RNA Isolation Kit with Manual - FAQs (AM1951)
4 product FAQs found
What is the difference between the RiboPure Blood Purification Kit (Cat. No. AM1928) and Mouse RiboPure Blood RNA Isolation kit (Cat. No. AM1951)?
The former (Cat. No. AM1928) is for isolation of RNA from human blood and the latter (Cat. No. AM1951) is for isolation of RNA from mouse and rat blood. Both kits have the same columns but differ in protocol and the wash 1 buffer, wash 2/3 buffer, and sodium acetate solutions. We do not recommend using the mouse kit for human blood samples. The mouse kit accommodates the difference in viscosity of mouse/rat vs. human blood and also the higher mouse and rat blood RNA yield. The mouse kit allows recovery of either total RNA containing small RNAs or total RNA depleted of small RNAs based on which protocol in the manual was followed. With the RiboPure Blood Purification Kit, using the standard protocol in the manual, you can isolate total RNA depleted of small RNAs, but with an alternate protocol, you can isolate total RNA containing small RNAs. Alternatively, if you need small RNAs alone, then you should follow the above alternate protocol for isolation of total RNAs containing small RNAs, and then follow Section IVA in the mirVana miRNA Isolation Kit manual (Cat. No AM1560).
What is the column cut-off size for the RiboPure RNA Purification Kits?
The column cut-off size is 200 nt. Using a RiboPure kit is not appropriate for miRNA recovery. For miRNA isolation, we would suggest using a mirVana kit, the TRIzol Plus RNA Purification Kit, or the PureLink RNA Micro Kit.
What are the differences in technology between the PureLink RNA Mini Kit and RiboPure RNA Purification Kits?
The PureLink RNA Mini Kit provides rapid column-based purification of total RNA, without organic lysis (phenol/chloroform). You can obtain up to 1,000 µg of purified RNA from a single extraction. The RiboPure RNA Purification Kits combine a phenol/guanidine thiocyanate solution with a glass-fiber filter purification method.
Do Tempus Blood RNA Tubes (Cat. No. 4342792) stabilize RNA similarly to RNAlater Stabilization Solution (Cat. No. AM7021)?
Both Tempus Blood RNA Tubes and RNAlater Stabilization Solution employ the same effective stabilization of mRNA expression profiles, and eliminate the need to isolate the RNA immediately after sample collection. However, there are some key differences:
- Components in each stabilization solution are different. The exact components and differences are proprietary.
- The RNAlater Stabilization Solution preserves RNA from degradation without lysing blood cells.
- Samples in RNAlater Stabilization Solution can be safely stored for up to 1 week at 25 degrees C, or up to 1 month at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.