UltraPure™ BSA (50 mg/mL), 1 x 50 mg - FAQs
UltraPure™ BSA (50 mg/mL), 1 x 50 mg - FAQs
View additional product information for UltraPure™ BSA (50 mg/mL) - FAQs (AM2618, AM2616)
1 product FAQs found
What is BSA and its function in PCR? What concentration of BSA do you recommend?
BSA, bovine serum albumin, is usually used in PCR as a stabilizer. A concentration of 160 µg/mL is typical. Gelatin may also be used as a stabilizer at 0.01% (w/v).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.