What should I consider when selecting a process density gauge?
When choosing a density gauge, consider:
- Process fluid (liquid or slurry) density range
- Pipe material type and diameter
- Operating and ambient temperature
- Required measurement range
- Accuracy and update time requirements
- Available plant communication protocols
- Access for operation, maintenance, calibration and verification
- Whether the application benefits from a non-intrusive measurement approach
For abrasive, corrosive, toxic, or difficult to access process lines, a non-contact nucleonic density gauge like the Thermo Scientific DDG3 Digital Density Gauge is designed to provide reliable, continuous measurement with minimal maintenance and no process interruption during installation.
How does the Thermo Scientific DDG3 Digital Density Gauge maintain long term measurement accuracy?
The Thermo Scientific DDG3 Digital Density Gauge incorporates high-performance microprocessor electronics, sensitive NaI scintillation detectors, automatic gain stabilisation and automatic source decay compensation. These features help maintain consistent measurement performance over the life of the radioactive source while helping reduce the need for frequent recalibration or maintenance.
Is the Thermo Scientific DDG3 Digital Density Gauge suitable for harsh industrial environments?
Yes. The Thermo Scientific DDG3 Digital Density Gauge is engineered for demanding process environments. Its controller (transmitter) features an IP66-rated enclosure, while the detector and source assemblies are built for reliable long-term operation in dusty, wet, and challenging industrial conditions. The system is designed for reliable operation in applications involving abrasive slurries, corrosive chemicals, and elevated process temperatures.
What communication protocols does the Thermo Scientific DDG3 Digital Density Gauge support?
The Thermo Scientific DDG3 Digital Density Gauge is designed for integration into modern industrial automation systems. Standard communication options include Modbus RTU and Modbus TCP. Optional protocols include HART, PROFINET, and EtherNet/IP, allowing straightforward integration with PLCs, DCS platforms, and plant control systems.
Can the Thermo Scientific DDG3 Digital Density Gauge measure percent solids as well as density?
Yes. The Thermo Scientific DDG3 Digital Density Gauge can be calibrated to measure process density and percent solids, depending on the application. Its 4-line display can be configured to show 3 readings from your choice of 5 measurands. These choices of measurands are also available for its two programmable current loop outputs and over serial and Ethernet protocols. This makes it especially valuable in plants where maintaining the correct solids concentration is critical for process quality, throughput, and energy efficiency.
What pipe sizes can the Thermo Scientific DDG3 Digital Density Gauge measure?
The standard Thermo Scientific DDG3 Digital Density Gauge system supports pipe diameters from 40 mm to 600 mm, with custom mounting hardware available for pipes ranging up to 1000 mm. This flexibility allows the system to be used across a wide range of industrial process applications.
Why choose a non-intrusive density gauge instead of an inline density meter?
A non-intrusive density gauge offers several operational advantages:
- No interruption of the production process during installation
- No process contamination risk
- No wetted parts exposed to abrasion or corrosion
- Lower maintenance requirements
- Suitable for hazardous, toxic, corrosive, or high temperature applications
- Reduced downtime because the instrument mounts outside the pipe
These advantages can help reduce lifetime operating costs.
Which industries benefit most from the Thermo Scientific DDG3 Digital Density Gauge?
The Thermo Scientific DDG3 Digital Density Gauge is widely used in industries where continuous density measurement improves process efficiency and product quality, including:
- Mining and mineral processing
- Coal preparation
- Chemical and petrochemical processing
- Food and beverage manufacturing
- Pulp and paper production
- Water and wastewater treatment
Its rugged, non-contact design makes it particularly well suited for challenging industrial environments.
How does a nucleonic density gauge measure density?
The Thermo Scientific DDG3 Digital Density Gauge uses gamma ray attenuation to determine the density of the process material inside a pipe. A sealed industrial radioactive source emits gamma rays through the pipe while a highly sensitive scintillation detector measures the amount of gamma radiation that reaches the detector. As the process density changes, the detected radiation changes inversely proportionally, allowing the system to calculate density or percent solids with high accuracy without contacting the process.
What is the Thermo Scientific DDG3 Digital Density Gauge?
The Thermo Scientific DDG3 Digital Density Gauge is a non-intrusive, non-contact process density measurement system designed to continuously measure the density or percent solids of liquids and slurries flowing through pipelines. Because it mounts externally to the pipe, it eliminates the need to cut into the process line, making it ideal for abrasive, corrosive, toxic, or elevated temperature applications.