NaCl (5 M), RNase-free, 100 mL - FAQs
NaCl (5 M), RNase-free, 100 mL - FAQs
View additional product information for NaCl (5 M), RNase-free - FAQs (AM9760G, AM9759)
1 product FAQs found
What can I do to prevent my DNA standards from smearing during electrophoresis?
Add 10 to 20 mM NaCl to the sample to maintain proper ionic strength.