4X Bolt™ LDS Sample Buffer
Invitrogen™

4X Bolt™ LDS Sample Buffer

Bolt LDS Sample Buffer (4X) is used to prepare protein samples for denaturing gel electrophoresis with Bis-Tris Plus gels. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
B0008250 mL
B000710 mL
Catalog number B0008
Price (USD)
181.65
Online exclusive
193.00
Save 11.35
Each
Quantity:
250 mL
Price (USD)
181.65
Online exclusive
193.00
Save 11.35
Each
Bolt LDS Sample Buffer (4X) is used to prepare protein samples for denaturing gel electrophoresis with Bis-Tris Plus gels. It contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

See all available buffers and reagents available for SDS-PAGE

Bolt LDS Sample Buffer contains Coomassie G250 and Phenol Red as tracking dyes instead of bromophenol blue. Coomassie G250 gives a sharp dye front with both MES and MOPS SDS running buffers and migrates much closer to the moving ion front than bromophenol blue. Bromophenol blue runs more slowly than some peptides with MES SDS Running Buffer. This ensures that small peptides do not run off the gels.

To use: Heat the sample in a 1X dilution (reduced or non-reduced) at 70°C for 10 minutes for optimal results.

Note: Bolt LDS Sample Buffer should be brought to room temperature (25°C) prior to use. It is a highly viscous and concentrated solution containing twice the amount of LDS compared to the amount of SDS in typical sample buffers. Bolt LDS Sample Buffer also contains a higher concentration glycerol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration4X
For Use With (Equipment)Bolt Mini Gel Tank
Gel CompatibilityBolt™ Bis-Tris Plus Gels
Quantity250 mL
Gel TypeBis-Tris Plus
Product LineBolt
Product TypeLDS Sample Buffer
Unit SizeEach
Contents & Storage
Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the shelf life for Bolt gels?

The shelf life for Bolt gels is similar to that for NuPAGE gels: 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.