D-Biotin, 10 mL - FAQs
D-Biotin, 10 mL - FAQs
View additional product information for D-Biotin - FAQs (B20656, B1595)
1 product FAQs found
How should I resolubilize D-biotin (B20656)?
D-biotin may be resolubilized in DMF (1-2 mg/mL), DMSO (49 mg/mL), ethanol (<1 mg/mL) or a buffer with a pH between >6 and <9. For long-term stability, D-biotin is stable in aqueous buffers at moderately acidic to neutral pH.