D-Biotin
Invitrogen™

D-Biotin

D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
B2065610 mL
B15951 g
카탈로그 번호 B20656
제품 가격(KRW)
191,000
온라인 행사
Ends: 30-Sep-2026
224,000
할인액 33,000 (15%)
Each
수량:
10 mL
제품 가격(KRW)
191,000
온라인 행사
Ends: 30-Sep-2026
224,000
할인액 33,000 (15%)
Each
D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins.
For Research Use Only. Not for use in diagnostic procedures.
사양
농도50mM
제품 유형Biotin (unconjugated)
수량10 mL
배송 조건Room Temperature
콘주게이트Unconjugated
형태Liquid
제품라인Invitrogen
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C).

자주 묻는 질문(FAQ)

How should I resolubilize D-biotin (B20656)?

D-biotin may be resolubilized in DMF (1-2 mg/mL), DMSO (49 mg/mL), ethanol (<1 mg/mL) or a buffer with a pH between >6 and <9. For long-term stability, D-biotin is stable in aqueous buffers at moderately acidic to neutral pH.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (2)

인용 및 참조 문헌
Abstract
Easily reversible desthiobiotin binding to streptavidin, avidin, and other biotin-binding proteins: uses for protein labeling, detection, and isolation.
Authors:Hirsch JD, Eslamizar L, Filanoski BJ, Malekzadeh N, Haugland RP, Beechem JM, Haugland RP
Journal:Anal Biochem
PubMed ID:12419349
'The high-affinity binding of biotin to avidin, streptavidin, and related proteins has been exploited for decades. However, a disadvantage of the biotin/biotin-binding protein interaction is that it is essentially irreversible under physiological conditions. Desthiobiotin is a biotin analogue that binds less tightly to biotin-binding proteins and is easily displaced by ... More
A SPECTROPHOTOMETRIC ASSAY FOR AVIDIN AND BIOTIN BASED ON BINDING OF DYES BY AVIDIN.
Authors:GREEN NM,
Journal:Biochem J
PubMed ID:14340040
A spectrophotometric assay for avidin and biotin based on binding of dyes by avidin.