BacLight™ RedoxSensor™ CTC Vitality Kit - FAQs
BacLight™ RedoxSensor™ CTC Vitality Kit - FAQs
View additional product information for BacLight™ RedoxSensor™ CTC Vitality Kit - FAQs (B34956)
1 product FAQs found
What bacterial parameters can I look at by flow cytometry?
You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).