Prepare stock solutions with confidence using fresh, single-use size anhydrous DMSO (order D12345)
<i>Bac</i>Light&trade; Red Bacterial Stain
Invitrogen™

BacLight™ Red Bacterial Stain

The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detectingRead more
Have Questions?
Catalog NumberQuantity
B3500120 x 50 μg
Catalog number B35001
Price (USD)
456.00
Each
Quantity:
20 x 50 μg
Price (USD)
456.00
Each
The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detecting and monitoring bacteria. These two dyes are not nucleic acid stains. Bacteria stained with the BacLight Green and BacLight Red bacterial stains exhibit bright green and red fluorescence (absorption/emission ∼480/516 and ∼581/644 nm, respectively), and can be resolved using the appropriate flow cytometric channels or fluorescence microscopy filters. The BacLight bacterial stains are compatible with formaldehyde or alcohol fixation methods.

View additional information about all microbiological analysis products.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeBacteria
Detection MethodFluorescence
Dye TypeOther Label(s) or Dye(s)
Emission Wavelength488
FormSolid
FormatTube(s), Slide(s)
Quantity20 x 50 μg
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineBacLight
Product TypeBacterial Stain
Unit SizeEach
Contents & Storage
Contains 20 vials of BacLight™ red bacterial stain (50 ug per vial). Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

What bacterial parameters can I look at by flow cytometry?

You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (2)

Citations & References
Abstract
Streptococcus pyogenes Biofilm Growth In Vitro and In Vivo and Its Role in Colonization, Virulence, and Genetic Exchange.
Authors:Marks LR, Mashburn-Warren L, Federle MJ, Hakansson AP,
Journal:
PubMed ID:24465015
'Background. Group A streptococcus (GAS) commonly colonizes the oropharynx and nonintact skin. However, colonization has been little studied and the role of biofilm formation is unclear, as biofilm experiments to date have not been conducted under conditions that mimic the host environment.Methods. In this study we grew GAS biofilms on ... More
Dual protective mechanisms of matrix metalloproteinases 2 and 9 in immune defense against Streptococcus pneumoniae.
Authors:Hong JS, Greenlee KJ, Pitchumani R, Lee SH, Song LZ, Shan M, Chang SH, Park PW, Dong C, Werb Z, Bidani A, Corry DB, Kheradmand F,
Journal:J Immunol
PubMed ID:21508260
A localized and effective innate immune response to pathogenic bacterial invasion is central to host survival. Identification of the critical local innate mediators of lung defense against such pathogens is essential for a complete understanding of the mechanism(s) underlying effective host defense. In an acute model of Streptococcus pneumoniae lung ... More