What is the 1X buffer composition of your TAE electrophoresis buffer (Cat. No. B49)?
The 1X buffer composition is as follows: 40 mM Tris, 20 mM acetic acid, 1mM EDTA.
Search
View additional product information for TAE Buffer (Tris-acetate-EDTA) (50X) - FAQs (B49)
1 product FAQs found